Assay Details
Target Gene Details
Entrez Gene ID: | 56949 |
Gene Name: | XPA binding protein 2 |
Gene Aliases: |
HCNP, HCRN, NTC90, SYF1 |
Location: |
Chr.19:7619525-7629553 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XAB2 | NM_020196.2 | NP_064581.2 | ||
AB026111.1 | BAB15807.1 | |||
AB033003.1 | BAA86491.1 | |||
AF226051.1 | AAF86951.1 | |||
AF258567.1 | AAG23770.1 | |||
AF272147.1 | AAQ14304.1 | |||
AK025858.1 | ||||
AK074035.1 | 8 | 2822 | BAB84861.1 | |
BC007208.1 | AAH07208.1 | |||
BC008778.1 | AAH08778.1 | |||
CR749864.1 | CAH18708.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953958 | Chr.19:7584715 - 7650814 on Build GRCh38 | Deletion | STXBP2 CAMSAP3 PET100 PCP2 MIR6792 XAB2 |
nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss | CLEC4G STXBP2 RETN CAMSAP3 FCER2 PET100 PCP2 MIR6792 XAB2 MCEMP1 TRAPPC5 |
nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain | CLEC4G ZNF358 EVI5L RETN INSR CAMSAP3 FCER2 PET100 PCP2 PNPLA6 MIR6792 MCOLN1 TRAPPC5 LOC107985283 STXBP2 LOC100128573 C19orf45 CD209 CLEC4GP1 CLEC4M XAB2 MCEMP1 PEX11G ARHGEF18 |
esv993190 | Chr.19:7615627 - 7629366 on Build GRCh38 | Deletion | CAMSAP3 MIR6792 XAB2 |
nsv833730 | Chr.19:7509505 - 7633597 on Build GRCh38 | Loss | ZNF358 CAMSAP3 PET100 PCP2 PNPLA6 MIR6792 MCOLN1 XAB2 |
esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain | MBD3L4 CLEC4G ZNF358 ADGRE1 ADGRE4P INSR MBD3L3 MCOLN1 TRAPPC5 MBD3L2 STXBP2 C19orf45 CLEC4GP1 CLEC4M PEX11G ZNF557 MBD3L5 RETN CAMSAP3 FCER2 PET100 PCP2 PNPLA6 MIR6792 LOC107985283 FLJ25758 LOC100128573 CD209 XAB2 LOC729458 MCEMP1 ARHGEF18 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |