Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | NP_005348.2 | ||
XM_005258937.3 | XP_005258994.1 | |||
XM_005258938.4 | XP_005258995.1 | |||
XM_006723218.2 | XP_006723281.1 | |||
AK292482.1 | ||||
BC070041.1 | AAH70041.1 | |||
U40002.1 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | ATP1A3 LOC101930071 LIPE-AS1 ZNF526 ARHGEF1 MEGF8 TMEM145 DEDD2 CNFN ZNF574 LOC390937 PRR19 MIR6797 CD79A POU2F2 RPS19 LIPE LOC100505585 CIC MIR4323 ERF GSK3A MIR8077 RABAC1 PAFAH1B3 GRIK5 LOC100505622 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | POU2F2 LOC101930071 LIPE CIC LIPE-AS1 ZNF526 ERF GSK3A MEGF8 TMEM145 CEACAM1 DEDD2 CXCL17 CNFN MIR8077 PAFAH1B3 PRR19 CEACAM8 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | ATP1A3 LOC101930071 LIPE-AS1 ZNF526 ARHGEF1 DMRTC2 MEGF8 CEACAM7 TMEM145 DEDD2 CNFN ZNF574 LOC390937 PRR19 MIR6797 CD79A POU2F2 RPS19 CEACAM4 LIPE LOC100505585 CIC MIR4323 ERF CEACAM3 CEACAM5 GSK3A MIR8077 RABAC1 PAFAH1B3 GRIK5 CEACAM6 LOC100505622 LYPD4 |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | PSG10P PSG8 PSG7 PSG2 LOC101930071 LIPE CIC PSG11 LIPE-AS1 ERF LOC100289650 PSG1 GSK3A MEGF8 TMEM145 CEACAM1 CXCL17 CNFN MIR8077 PAFAH1B3 PRR19 PSG6 CEACAM8 PSG3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |