Assay Details
Target Gene Details
Entrez Gene ID: | 100506033 |
Gene Name: | PTOV1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:49838645-49851676 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1-AS1 | NR_040037.1 | |||
AK092213.1 | 1 | 1929 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | TSKS RUVBL2 RPL18 SPACA4 LMTK3 LIN7B FUT2 PTOV1-AS1 HRC NUP62 SNAR-C5 RRAS SNAR-A8 EMP3 MIR4324 SNAR-B1 PTH2 DBP KDELR1 ZNF473 CGB7 SNAR-G1 RCN3 HSD17B14 CA11 CGB8 PLEKHA4 SNAR-C2 SNAR-A9 PRR12 GRIN2D SYNGR4 LOC100287477 ADM5 NTN5 DKKL1 SLC17A7 BSPH1 FLT3LG SNAR-G2 SLC6A16 GFY SIGLEC16 C19orf68 BAX DHDH CARD8-AS1 FLJ26850 FTL MIR6800 MIR4750 SNAR-C3 PLA2G4C SNAR-A6 MIR5088 MIR4751 SPHK2 SNORD35B PRRG2 LIG1 GRWD1 LHB CARD8 SEC1P RPL13A PPFIA3 AKT1S1 TRPM4 C19orf73 RPS11 CYTH2 CGB5 ELSPBP1 IRF3 TBC1D17 LOC101928295 SNORD33 TEAD2 ALDH16A1 NTF4 CGB1 PTOV1 CD37 BCAT2 KCNJ14 SULT2B1 AP2A1 MIR6799 IL4I1 CCDC155 SNAR-A4 CCDC114 SNAR-A14 IZUMO1 MAMSTR SNORD34 GYS1 SNORD35A NOSIP FCGRT LOC105372430 PIH1D1 CGB2 SNRNP70 MIR6798 FUZ FAM83E LOC105447645 SNAR-A7 TULP2 NUCB1 CGB3 SNAR-A3 FUT1 PTOV1-AS2 TMEM143 CABP5 MIR4749 SNAR-C1 CPT1C SNAR-A2 VRK3 SIGLEC11 SNAR-B2 SNAR-A1 KCNA7 SNORD32A SNAR-A10 ZNF114 SCAF1 SNAR-A5 ATF5 SNAR-A12 PPP1R15A PLA2G4C-AS1 LOC101059948 SNAR-A13 MED25 PRMT1 SNAR-A11 FGF21 PNKP MIR150 SNAR-D SNAR-C4 NUCB1-AS1 BCL2L12 RASIP1 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | MIR5088 TSKS SCAF1 PTOV1-AS1 IRF3 MIR4749 MED25 PRR12 PRMT1 RRAS MIR6800 ADM5 CPT1C PTOV1 FUZ BCL2L12 PTOV1-AS2 AP2A1 MIR6799 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | AKT1S1 TSKS MIR4751 IL4I1 ATF5 PTOV1-AS1 TBC1D17 MIR4749 MED25 NUP62 MIR6800 MIR4750 PNKP PTOV1 FUZ PTOV1-AS2 AP2A1 MIR6799 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | AKT1S1 TSKS PTOV1-AS1 IRF3 TBC1D17 MIR4749 NUP62 RRAS MIR6800 CPT1C MIR4750 PTOV1 AP2A1 MIR6799 MIR5088 MIR4751 IL4I1 SCAF1 ATF5 PRRG2 MED25 PRR12 PRMT1 ADM5 NOSIP PNKP FUZ BCL2L12 PTOV1-AS2 |
dgv54n68 | Chr.19:49848512 - 50008788 on Build GRCh38 | Loss | AKT1S1 SIGLEC16 MIR4751 IL4I1 ATF5 PTOV1-AS1 TBC1D17 MIR4749 NUP62 MIR4750 PNKP PTOV1 VRK3 SIGLEC11 PTOV1-AS2 |