Assay Details
Target Gene Details
Entrez Gene ID: | 126308 |
Gene Name: | MOB kinase activator 3A |
Gene Aliases: |
MOB1C, MOBKL2A, moblak |
Location: |
Chr.19:2071036-2096502 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOB3A | NM_130807.2 | NP_570719.1 | ||
XM_011527683.2 | XP_011525985.1 | |||
AB061793.1 | BAB84554.1 | |||
AK095471.1 | ||||
AL832474.1 | ||||
BC015049.1 | AAH15049.1 | |||
CR627409.1 | CAH10498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss | ONECUT3 MKNK2 LOC100288123 ADAT3 SCAMP4 CSNK1G2-AS1 BTBD2 ABHD17A REXO1 KLF16 MIR1909 ATP8B3 IZUMO4 AP3D1 MOB3A CSNK1G2 |
nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss | MKNK2 LOC100288123 ADAT3 SCAMP4 CSNK1G2-AS1 BTBD2 ABHD17A REXO1 KLF16 MIR1909 ATP8B3 IZUMO4 MOB3A CSNK1G2 |
esv3583203 | Chr.19:2044317 - 2080698 on Build GRCh38 | Loss | MKNK2 MOB3A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |