Assay Details
Target Gene Details
Entrez Gene ID: | 55723 |
Gene Name: | anti-silencing function 1B histone chaperone |
Gene Aliases: |
CIA-II |
Location: |
Chr.19:14119509-14136628 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASF1B | NM_018154.2 | NP_060624.1 | ||
AB104486.1 | BAC87709.1 | |||
AF279307.1 | AAK82973.1 | |||
AK001288.1 | BAA91602.1 | |||
AK001466.1 | BAA91708.1 | |||
AK223080.1 | BAD96800.1 | |||
AK302134.1 | ||||
BC007726.2 | AAH07726.1 | |||
BC010014.2 | AAH10014.1 | |||
BC036521.1 | AAH36521.1 | |||
CN426445.1 | ||||
CR457235.1 | CAG33516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833761 | Chr.19:13998659 - 14136778 on Build GRCh38 | Loss | PALM3 C19orf67 MIR1199 IL27RA SAMD1 RFX1 RLN3 MISP3 ASF1B PRKACA |
nsv953980 | Chr.19:14082589 - 14210288 on Build GRCh38 | Deletion | C19orf67 LOC100507373 SAMD1 ADGRL1 ASF1B PRKACA |
dgv49n68 | Chr.19:14066502 - 14274131 on Build GRCh38 | Loss | C19orf67 MIR1199 LOC100507373 SAMD1 ADGRL1 MISP3 ASF1B PRKACA |
nsv828452 | Chr.19:14059742 - 14148154 on Build GRCh38 | Loss | PALM3 C19orf67 MIR1199 LOC100507373 SAMD1 ADGRL1 MISP3 ASF1B PRKACA |
nsv578642 | Chr.19:14028192 - 14187630 on Build GRCh38 | Gain | PALM3 C19orf67 MIR1199 IL27RA LOC100507373 SAMD1 ADGRL1 RLN3 MISP3 ASF1B PRKACA |
nsv578663 | Chr.19:14111697 - 14226588 on Build GRCh38 | Loss | LOC100507373 ADGRL1 ASF1B PRKACA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |