Assay Details
Target Gene Details
Entrez Gene ID: | 360200 |
Gene Name: | transmembrane protease, serine 9 |
Gene Aliases: |
- |
Location: |
Chr.19:2385254-2426261 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS9 | NM_182973.1 | NP_892018.1 | ||
XM_011527978.2 | XP_011526280.1 | |||
AB109390.1 | ||||
AJ488946.1 | CAD35758.1 | |||
AJ488947.1 | CAD35759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160562 | Chr.19:2328434 - 2460264 on Build GRCh38 | Deletion | TMPRSS9 MIR7108 LSM7 LMNB2 SPPL2B TIMM13 LOC101928602 |
esv3583208 | Chr.19:2389734 - 2409683 on Build GRCh38 | Loss | TMPRSS9 |
nsv1064930 | Chr.19:2380093 - 2592252 on Build GRCh38 | Gain | LOC101929097 TMPRSS9 MIR7108 GADD45B LMNB2 TIMM13 LOC101928602 GNG7 |
nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion | LOC101929097 LINGO3 TMPRSS9 MIR7108 LSM7 GADD45B LMNB2 SPPL2B TIMM13 LOC101928602 GNG7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |