Assay Details
Target Gene Details
Entrez Gene ID: | 1830 |
Gene Name: | desmoglein 3 |
Gene Aliases: |
CDHF6, PVA |
Location: |
Chr.18:31447769-31478702 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DSG3 | NM_001944.2 | NP_001935.2 | ||
XM_011525850.1 | XP_011524152.1 | |||
AK290367.1 | ||||
DA458544.1 | ||||
M76482.1 | AAA60230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv438278 | Chr.18:31453177 - 31454341 on Build GRCh38 | Loss |
![]() |
esv2656541 | Chr.18:31452541 - 31454012 on Build GRCh38 | Deletion |
![]() |
nsv437177 | Chr.18:31449805 - 31459320 on Build GRCh38 | Loss |
![]() |
dgv3328n100 | Chr.18:31385807 - 31473928 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3642128 | Chr.18:31452459 - 31454052 on Build GRCh38 | Loss |
![]() |
dgv975e212 | Chr.18:31385113 - 31471686 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2751776 | Chr.18:31390184 - 31477756 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv524890 | Chr.18:31387447 - 31476502 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv833615 | Chr.18:31428433 - 31613013 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv5882n54 | Chr.18:31389081 - 31476502 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)