Assay Details
Target Gene Details
Entrez Gene ID: | 91768 |
Gene Name: | Cdk5 and Abl enzyme substrate 1 |
Gene Aliases: |
CABL1, CABLES, HsT2563, IK3-1 |
Location: |
Chr.18:23134564-23260470 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABLES1 | NM_001100619.2 | NP_001094089.1 | ||
NM_001256438.1 | NP_001243367.1 | |||
NM_138375.2 | NP_612384.1 | |||
NR_023359.1 | ||||
AF348525.1 | AAL83906.1 | |||
AK093243.1 | BAC04107.1 | |||
AK296407.1 | ||||
AK304880.1 | ||||
BC004124.2 | AAH04124.2 | |||
BC037218.1 | AAH37218.1 | |||
EF028204.1 |
Target Gene Details
Entrez Gene ID: | 85019 |
Gene Name: | transmembrane protein 241 |
Gene Aliases: |
C18orf45, hVVT |
Location: |
Chr.18:23237252-23437969 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM241 | XM_017026043.1 | XP_016881532.1 | ||
XM_017026046.1 | XP_016881535.1 | |||
XM_017026047.1 | XP_016881536.1 | |||
XM_017026048.1 | XP_016881537.1 | |||
AK294916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758716 | Chr.18:23190468 - 23347650 on Build GRCh38 | Loss | TMEM241 CABLES1 |
esv2761994 | Chr.18:23241557 - 23257139 on Build GRCh38 | Gain | TMEM241 CABLES1 |
esv32657 | Chr.18:23203968 - 23353903 on Build GRCh38 | Loss | TMEM241 CABLES1 |
nsv458033 | Chr.18:23220751 - 23290196 on Build GRCh38 | Loss | TMEM241 CABLES1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |