Assay Details
Target Gene Details
Entrez Gene ID: | 84064 |
Gene Name: | haloacid dehalogenase like hydrolase domain containing 2 |
Gene Aliases: |
3110052N05Rik, HEL-S-301 |
Location: |
Chr.18:47107402-47150520 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HDHD2 | NM_001318765.1 | NP_001305694.1 | ||
NM_032124.4 | NP_115500.1 | |||
XM_005258371.4 | XP_005258428.1 | |||
XM_006722557.2 | XP_006722620.1 | |||
XM_011526227.2 | XP_011524529.1 | |||
XM_011526228.2 | XP_011524530.1 | |||
XM_017026039.1 | XP_016881528.1 | |||
XM_017026040.1 | XP_016881529.1 | |||
AK054590.1 | BAB70768.1 | |||
AK292098.1 | ||||
AL136681.1 | CAB66616.1 | |||
BC011894.2 | AAH11894.1 | |||
BC033031.1 | AAH33031.1 | |||
BC038100.2 | AAH38100.1 | |||
BE246996.1 | ||||
FJ224322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3642451 | Chr.18:47089642 - 47219390 on Build GRCh38 | Gain | SKOR2 HDHD2 KATNAL2 IER3IP1 |
nsv576819 | Chr.18:47075878 - 47151256 on Build GRCh38 | Loss | HDHD2 KATNAL2 |
nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss | TCEB3CL2 TCEB3B LOC107983955 SKOR2 HDHD2 PIAS2 KATNAL2 ST8SIA5 TCEB3CL TCEB3C IER3IP1 |
nsv833640 | Chr.18:47025905 - 47188501 on Build GRCh38 | Loss | TCEB3B HDHD2 KATNAL2 TCEB3C IER3IP1 |
esv3642452 | Chr.18:47093035 - 47161369 on Build GRCh38 | Gain | HDHD2 KATNAL2 IER3IP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |