Assay Details
Target Gene Details
Entrez Gene ID: | 9625 |
Gene Name: | apoptosis associated tyrosine kinase |
Gene Aliases: |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
Location: |
Chr.17:81117295-81166072 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK | NM_001080395.2 | NP_001073864.2 | ||
NM_004920.2 | NP_004911.2 | |||
XM_006722193.2 | XP_006722256.1 | |||
XM_006722194.2 | XP_006722257.1 | |||
XM_006722195.2 | XP_006722258.1 | |||
XM_006722196.2 | XP_006722259.1 | |||
XM_011525509.2 | XP_011523811.1 | |||
XM_011525510.2 | XP_011523812.1 | |||
XM_011525511.2 | XP_011523813.1 | |||
XM_017025421.1 | XP_016880910.1 | |||
XM_017025422.1 | XP_016880911.1 | |||
AB014541.2 | ||||
AK094931.1 | ||||
AK131395.1 | BAD18544.1 | |||
AK131529.1 | BAD18667.1 | |||
BC047378.1 | AAH47378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | SLC25A10 MIR657 NPLOC4 MRPL12 MIR6786 TMEM105 LOC107985021 BAHCC1 LOC105371925 CHMP6 LOC400627 BAIAP2 AATK-AS1 MIR338 LINC00482 RPTOR MIR1250 FAAP100 SLC38A10 ACTG1 TEPSIN MIR4740 ARL16 LOC107985022 C17orf89 LOC100129503 AATK PDE6G BAIAP2-AS1 LOC101928855 LOC100130370 CCDC137 CEP131 HGS FSCN2 MIR3186 OXLD1 MIR3065 LOC105371921 TSPAN10 |
esv2716426 | Chr.17:81119453 - 81119928 on Build GRCh38 | Deletion | AATK |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | MIR657 MIR338 AATK MIR1250 BAIAP2 CEP131 MIR3065 AATK-AS1 |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | MIR657 MIR338 AATK MIR1250 BAIAP2-AS1 BAIAP2 MIR3065 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | MIR657 MIR338 LINC00482 MIR1250 SLC38A10 TEPSIN TMEM105 LOC105371925 C17orf89 AATK BAIAP2-AS1 BAIAP2 CEP131 MIR3065 AATK-AS1 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR657 MIR338 AATK MIR1250 BAIAP2-AS1 BAIAP2 CEP131 MIR3065 AATK-AS1 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | MIR657 MIR338 AATK MIR1250 BAIAP2 MIR3065 AATK-AS1 |
nsv523417 | Chr.17:81115790 - 81164271 on Build GRCh38 | Gain | MIR657 MIR338 AATK MIR1250 BAIAP2 MIR3065 |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | MIR657 MIR338 AATK MIR1250 BAIAP2-AS1 BAIAP2 MIR3065 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |