Assay Details
Target Gene Details
Entrez Gene ID: | 125206 |
Gene Name: | solute carrier family 5 member 10 |
Gene Aliases: |
SGLT-5, SGLT5 |
Location: |
Chr.17:18950676-19020691 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC5A10 | NM_001042450.2 | NP_001035915.1 | ||
NM_001270648.1 | NP_001257577.1 | |||
NM_001270649.1 | NP_001257578.1 | |||
NM_001282417.1 | NP_001269346.1 | |||
NM_152351.4 | NP_689564.3 | |||
XM_011523652.1 | XP_011521954.1 | |||
XM_011523653.2 | XP_011521955.1 | |||
XM_017024191.1 | XP_016879680.1 | |||
AK057946.1 | BAB71619.1 | |||
AK298345.1 | ||||
BC034380.1 | ||||
BC039868.1 | AAH39868.1 | |||
BC062617.1 | AAH62617.1 | |||
BP332461.1 | ||||
DR003789.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574552 | Chr.17:18926974 - 18987449 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1055999 | Chr.17:18695458 - 18973671 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv33572 | Chr.17:18919047 - 19291177 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758678 | Chr.17:18927303 - 19288734 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1057803 | Chr.17:18561260 - 19019420 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv953842 | Chr.17:18947088 - 19049187 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv523721 | Chr.17:18659407 - 19014011 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833391 | Chr.17:18906887 - 19238887 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv565e214 | Chr.17:18920166 - 19019319 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv428336 | Chr.17:18927239 - 19288734 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833390 | Chr.17:18786912 - 18976381 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv9504 | Chr.17:18389579 - 19238707 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv5491n54 | Chr.17:18921239 - 19014011 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv470581 | Chr.17:18947699 - 19014011 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)