Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3554876 | Chr.17:81169201 - 81174848 on Build GRCh38 | Deletion | AATK-AS1 |
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | TEPSIN FSCN2 NPLOC4 MIR1250 LOC400627 PDE6G HGS FAAP100 SLC25A10 OXLD1 CEP131 BAIAP2-AS1 LOC101928855 ACTG1 LOC107985021 AATK-AS1 ARL16 MRPL12 LOC100130370 CHMP6 LOC105371921 BAHCC1 SLC38A10 LOC107985022 MIR6786 BAIAP2 MIR657 MIR3186 MIR3065 CCDC137 TSPAN10 LOC105371925 MIR338 LINC00482 LOC100129503 RPTOR MIR4740 C17orf89 AATK TMEM105 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | CEP131 AATK-AS1 AATK |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | STRA13 ZNF750 TBCD TEPSIN FSCN2 DCXR B3GNTL1 MYADML2 NPLOC4 PYCR1 FN3KRP NOTUM FAAP100 CEP131 NPB FOXK2 ACTG1 FASN MRPL12 MAFG-AS1 LOC100130370 PCYT2 BAHCC1 MIR6786 MIR4525 LRRC45 UTS2R ALYREF TSPAN10 NARF ARHGDIA LINC00482 ANAPC11 C17orf89 CCDC57 LOC101929552 MAFG GCGR LOC105376790 DUS1L PPP1R27 RAB40B LOC105376791 TEX19 LOC105371944 PDE6G HGS SLC25A10 SLC16A3 CD7 OXLD1 FN3K P4HB AATK-AS1 WDR45B ARL16 LOC101929511 CSNK1D RAC3 ASPSCR1 GPS1 SLC38A10 MIR6787 MIR3186 CCDC137 SECTM1 C17orf62 SIRT7 LOC105371925 SNORD134 MCRIP1 MIR4740 OGFOD3 HEXDC AATK TMEM105 RFNG |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | CEP131 LOC105371925 TEPSIN SLC38A10 C17orf89 AATK-AS1 AATK |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | CEP131 MIR338 AATK-AS1 BAIAP2 MIR657 AATK MIR1250 MIR3065 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | TEPSIN SLC38A10 BAIAP2 MIR657 MIR1250 MIR3065 CEP131 BAIAP2-AS1 LOC105371925 MIR338 LINC00482 C17orf89 AATK-AS1 AATK TMEM105 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | CEP131 BAIAP2-AS1 MIR338 AATK-AS1 BAIAP2 MIR657 AATK MIR1250 MIR3065 |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | CEP131 LOC105371925 TEPSIN SLC38A10 C17orf89 AATK-AS1 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | LOC100130370 BAHCC1 TEPSIN FSCN2 SLC38A10 NPLOC4 MIR3186 FAAP100 CEP131 LOC105371925 LINC00482 ACTG1 MIR4740 C17orf89 AATK-AS1 AATK TMEM105 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | MIR338 AATK-AS1 BAIAP2 MIR657 AATK MIR1250 MIR3065 |