Assay Details
Target Gene Details
Entrez Gene ID: | 10871 |
Gene Name: | CD300c molecule |
Gene Aliases: |
CLM-6, CMRF-35, CMRF-35A, CMRF35, CMRF35-A1, CMRF35A, CMRF35A1, IGSF16, LIR |
Location: |
Chr.17:74534359-74546171 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD300C | XM_017024033.1 | XP_016879522.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12e196 | Chr.17:73675043 - 74565539 on Build GRCh38 | Duplication | CD300C CD300A MGC16275 CD300LB DNAI2 RPL38 LINC00469 BTBD17 GPRC5C LOC100134391 LOC400620 GPR142 TTYH2 KIF19 LOC100130520 |
nsv470600 | Chr.17:74466882 - 74550979 on Build GRCh38 | Loss | CD300C CD300LB CD300A |
esv2761978 | Chr.17:73835799 - 74684915 on Build GRCh38 | Gain | CD300C CD300A CD300E MGC16275 C17orf77 CD300LB DNAI2 RPL38 CD300LD BTBD17 RAB37 GPRC5C GPR142 TTYH2 KIF19 LOC101928343 LOC100130520 |
esv3641230 | Chr.17:73831787 - 74694861 on Build GRCh38 | Gain | CD300C CD300A CD300E MGC16275 C17orf77 CD300LB DNAI2 RPL38 CD300LF CD300LD BTBD17 RAB37 GPRC5C GPR142 TTYH2 KIF19 LOC101928343 LOC100130520 |
dgv3282n100 | Chr.17:73814343 - 74719597 on Build GRCh38 | Gain | CD300C CD300A CD300E MGC16275 C17orf77 CD300LB DNAI2 RPL38 LINC00469 CD300LF CD300LD BTBD17 RAB37 GPRC5C GPR142 TTYH2 KIF19 LOC101928343 LOC100130520 |
esv33995 | Chr.17:74261053 - 74732093 on Build GRCh38 | Loss | CD300C CD300A CD300E C17orf77 CD300LB DNAI2 CD300LF CD300LD BTBD17 RAB37 GPRC5C GPR142 TTYH2 KIF19 LOC101928343 LOC100130520 |
nsv517569 | Chr.17:73837862 - 74685802 on Build GRCh38 | Gain+Loss | CD300C CD300A CD300E MGC16275 C17orf77 CD300LB DNAI2 RPL38 CD300LD BTBD17 RAB37 GPRC5C GPR142 TTYH2 KIF19 LOC101928343 LOC100130520 |
dgv5768n54 | Chr.17:73826016 - 74706775 on Build GRCh38 | Gain | CD300C CD300A CD300E MGC16275 C17orf77 CD300LB DNAI2 RPL38 LINC00469 CD300LF CD300LD BTBD17 RAB37 GPRC5C GPR142 TTYH2 KIF19 LOC101928343 LOC100130520 |
esv3573820 | Chr.17:73817932 - 74694378 on Build GRCh38 | Gain | CD300C CD300A CD300E MGC16275 C17orf77 CD300LB DNAI2 RPL38 LINC00469 CD300LF CD300LD BTBD17 RAB37 GPRC5C GPR142 TTYH2 KIF19 LOC101928343 LOC100130520 |
esv3641244 | Chr.17:74527395 - 74686883 on Build GRCh38 | Gain | CD300C C17orf77 CD300LB CD300LD RAB37 CD300E LOC101928343 LOC100130520 |
dgv5774n54 | Chr.17:74428381 - 74673021 on Build GRCh38 | Gain | CD300C C17orf77 CD300LB CD300LD RAB37 GPRC5C CD300A CD300E LOC101928343 LOC100130520 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |