Assay Details
Target Gene Details
Entrez Gene ID: | 105371907 |
Gene Name: | uncharacterized LOC105371907 |
Gene Aliases: |
- |
Location: |
Chr.17:77273784-77281897 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371907 | NR_136503.1 | |||
CB529354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833552 | Chr.17:77226556 - 77413897 on Build GRCh38 | Loss | SEPT9 LOC105371905 LOC105371907 LACAT1 MIR4316 |
dgv3284n100 | Chr.17:77118623 - 77628815 on Build GRCh38 | Gain | SEPT9 LOC100507351 LOC105371905 SEC14L1 LOC105371907 LACAT1 MIR4316 LOC400622 |
esv34174 | Chr.17:77098835 - 77357976 on Build GRCh38 | Loss | SEPT9 SEC14L1 LOC105371907 LACAT1 |
esv2758704 | Chr.17:77130826 - 77299326 on Build GRCh38 | Gain | SEPT9 SEC14L1 LOC105371907 LACAT1 |
dgv5787n54 | Chr.17:77270084 - 77281555 on Build GRCh38 | Loss | SEPT9 LOC105371907 LACAT1 |
nsv833551 | Chr.17:77138921 - 77309684 on Build GRCh38 | Gain | SEPT9 SEC14L1 LOC105371907 LACAT1 |
esv992724 | Chr.17:77267924 - 77281537 on Build GRCh38 | Deletion | SEPT9 LOC105371907 LACAT1 |
nsv576077 | Chr.17:77264304 - 77297220 on Build GRCh38 | Loss | SEPT9 LOC105371907 LACAT1 |
nsv517339 | Chr.17:77270169 - 77283227 on Build GRCh38 | Loss | SEPT9 LOC105371907 LACAT1 |