Assay Details
Target Gene Details
Entrez Gene ID: | 100874058 |
Gene Name: | COX10 antisense RNA 1 |
Gene Aliases: |
COX10-AS, COX10AS |
Location: |
Chr.17:14029292-14069458 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX10-AS1 | NR_049718.1 | |||
AK123100.1 | ||||
BI760398.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758675 | Chr.17:13977924 - 14225239 on Build GRCh38 | Loss | COX10 COX10-AS1 CDRT15P1 |
esv3640012 | Chr.17:14037121 - 14081238 on Build GRCh38 | Loss | COX10 COX10-AS1 |
nsv1056267 | Chr.17:13835722 - 14236763 on Build GRCh38 | Gain | LOC100506974 CDRT15 COX10 COX10-AS1 CDRT15P1 |
nsv574377 | Chr.17:13548048 - 14439233 on Build GRCh38 | Gain | HS3ST3A1 LOC100506974 MGC12916 HS3ST3B1 CDRT15 COX10 COX10-AS1 CDRT15P1 |
nsv1055511 | Chr.17:14042329 - 14191370 on Build GRCh38 | Loss | COX10 COX10-AS1 |