Assay Details
Target Gene Details
Entrez Gene ID: | 10052 |
Gene Name: | gap junction protein gamma 1 |
Gene Aliases: |
CX45, GJA7 |
Location: |
Chr.17:44794988-44830811 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GJC1 | NM_001080383.1 | NP_001073852.1 | ||
NM_005497.3 | NP_005488.2 | |||
XM_005256920.1 | XP_005256977.1 | |||
CN299428.1 | ||||
DA845933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833456 | Chr.17:44639788 - 44825873 on Build GRCh38 | Gain+Loss | LINC01180 DBF4B ADAM11 CCDC43 MEIOC GJC1 |
nsv828021 | Chr.17:44786920 - 44829084 on Build GRCh38 | Gain | GJC1 |
nsv523493 | Chr.17:44815427 - 44843561 on Build GRCh38 | Loss | GJC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |