Assay Details
Target Gene Details
Entrez Gene ID: | 105371907 |
Gene Name: | uncharacterized LOC105371907 |
Gene Aliases: |
- |
Location: |
Chr.17:77273784-77281897 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371907 | NR_136503.1 | 2 | 374 | |
CB529354.1 | 2 | 336 | ||
CD370373.1 | 2 | 691 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3554814 | Chr.17:77267106 - 77277918 on Build GRCh38 | Deletion |
![]() ![]() |
dgv3284n100 | Chr.17:77118623 - 77628815 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv34174 | Chr.17:77098835 - 77357976 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
dgv568e199 | Chr.17:77267821 - 77277205 on Build GRCh38 | Deletion |
![]() ![]() |
nsv1143863 | Chr.17:77267918 - 77277618 on Build GRCh38 | Deletion |
![]() ![]() |
nsv576077 | Chr.17:77264304 - 77297220 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3641310 | Chr.17:77269394 - 77276527 on Build GRCh38 | Loss |
![]() ![]() |
esv1007977 | Chr.17:77269552 - 77276494 on Build GRCh38 | Loss |
![]() ![]() |
nsv517339 | Chr.17:77270169 - 77283227 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv833552 | Chr.17:77226556 - 77413897 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2758704 | Chr.17:77130826 - 77299326 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv5787n54 | Chr.17:77270084 - 77281555 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv833551 | Chr.17:77138921 - 77309684 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv576087 | Chr.17:77272257 - 77276431 on Build GRCh38 | Loss |
![]() ![]() |
esv992724 | Chr.17:77267924 - 77281537 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)