Assay Details
Target Gene Details
Entrez Gene ID: | 55890 |
Gene Name: | G protein-coupled receptor class C group 5 member C |
Gene Aliases: |
RAIG-3, RAIG3 |
Location: |
Chr.17:74431341-74451658 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPRC5C | NM_018653.3 | NP_061123.3 | ||
NM_022036.2 | NP_071319.2 | |||
XM_005257512.1 | XP_005257569.1 | |||
XM_005257514.1 | XP_005257571.1 | |||
XM_017024855.1 | XP_016880344.1 | |||
XM_017024856.1 | XP_016880345.1 | |||
AB451470.1 | ||||
AF207989.1 | AAF72870.1 | |||
AJ276102.1 | CAC00633.1 | |||
AK000249.1 | BAA91030.1 | |||
AK075397.1 | BAC11595.1 | |||
AK126429.1 | 1 | 276 | ||
AK131210.1 | ||||
AK222909.1 | ||||
BC004925.1 | AAH04925.1 | |||
BC016860.1 | AAH16860.2 | |||
BC110848.1 | AAI10849.1 | |||
BC125080.1 | ||||
BC125081.1 | ||||
BT007435.1 | AAP36103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12e196 | Chr.17:73675043 - 74565539 on Build GRCh38 | Duplication | LINC00469 BTBD17 GPRC5C CD300LB KIF19 CD300A TTYH2 DNAI2 LOC100134391 GPR142 LOC400620 MGC16275 RPL38 CD300C LOC100130520 |
esv3641237 | Chr.17:74241755 - 74451924 on Build GRCh38 | Gain | GPR142 BTBD17 GPRC5C KIF19 TTYH2 DNAI2 |
esv2761978 | Chr.17:73835799 - 74684915 on Build GRCh38 | Gain | BTBD17 GPRC5C CD300LB KIF19 LOC101928343 CD300A TTYH2 DNAI2 CD300E RAB37 C17orf77 GPR142 MGC16275 RPL38 CD300LD CD300C LOC100130520 |
esv3641230 | Chr.17:73831787 - 74694861 on Build GRCh38 | Gain | CD300LF BTBD17 GPRC5C CD300LB KIF19 LOC101928343 CD300A TTYH2 DNAI2 CD300E RAB37 C17orf77 GPR142 MGC16275 RPL38 CD300LD CD300C LOC100130520 |
dgv3282n100 | Chr.17:73814343 - 74719597 on Build GRCh38 | Gain | CD300LF LINC00469 BTBD17 GPRC5C CD300LB KIF19 LOC101928343 CD300A TTYH2 DNAI2 CD300E RAB37 C17orf77 GPR142 MGC16275 RPL38 CD300LD CD300C LOC100130520 |
esv33995 | Chr.17:74261053 - 74732093 on Build GRCh38 | Loss | CD300LF BTBD17 GPRC5C CD300LB KIF19 LOC101928343 CD300A TTYH2 DNAI2 CD300E RAB37 C17orf77 GPR142 CD300LD CD300C LOC100130520 |
nsv517569 | Chr.17:73837862 - 74685802 on Build GRCh38 | Gain+Loss | BTBD17 GPRC5C CD300LB KIF19 LOC101928343 CD300A TTYH2 DNAI2 CD300E RAB37 C17orf77 GPR142 MGC16275 RPL38 CD300LD CD300C LOC100130520 |
dgv5768n54 | Chr.17:73826016 - 74706775 on Build GRCh38 | Gain | CD300LF LINC00469 BTBD17 GPRC5C CD300LB KIF19 LOC101928343 CD300A TTYH2 DNAI2 CD300E RAB37 C17orf77 GPR142 MGC16275 RPL38 CD300LD CD300C LOC100130520 |
nsv952366 | Chr.17:74429862 - 74456261 on Build GRCh38 | Deletion | GPRC5C |
esv3573820 | Chr.17:73817932 - 74694378 on Build GRCh38 | Gain | CD300LF LINC00469 BTBD17 GPRC5C CD300LB KIF19 LOC101928343 CD300A TTYH2 DNAI2 CD300E RAB37 C17orf77 GPR142 MGC16275 RPL38 CD300LD CD300C LOC100130520 |
dgv5774n54 | Chr.17:74428381 - 74673021 on Build GRCh38 | Gain | C17orf77 GPRC5C CD300LB CD300LD LOC101928343 CD300A CD300C LOC100130520 CD300E RAB37 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |