Assay Details
Target Gene Details
Entrez Gene ID: | 79701 |
Gene Name: | 2-oxoglutarate and iron dependent oxygenase domain containing 3 |
Gene Aliases: |
C17orf101 |
Location: |
Chr.17:82389210-82418637 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OGFOD3 | NM_024648.2 | NP_078924.1 | ||
NM_175902.4 | NP_787098.3 | |||
NR_033265.1 | ||||
AK025875.1 | BAB15267.1 | |||
AK311057.1 | ||||
BC008897.1 | AAH08897.1 | |||
BC023602.2 | AAH23602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059334 | Chr.17:82277007 - 82411683 on Build GRCh38 | Gain | SECTM1 UTS2R OGFOD3 LOC101929511 TEX19 LOC105376790 CD7 |
nsv833573 | Chr.17:82275053 - 82406183 on Build GRCh38 | Loss | SECTM1 UTS2R OGFOD3 LOC101929511 TEX19 LOC105376790 CD7 |
esv2760445 | Chr.17:82267669 - 82421456 on Build GRCh38 | Gain | SECTM1 HEXDC UTS2R OGFOD3 CSNK1D LOC101929511 TEX19 LOC105376790 CD7 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | LOC105376791 SLC16A3 GCGR P4HB FOXK2 MIR6786 AATK-AS1 NARF CEP131 LOC105371925 MIR6787 CCDC57 TEX19 SNORD134 PPP1R27 ACTG1 SLC38A10 OXLD1 DCXR RFNG ZNF750 FN3KRP C17orf62 PCYT2 HEXDC OGFOD3 CCDC137 TMEM105 LOC101929511 SIRT7 NPLOC4 LOC105376790 GPS1 B3GNTL1 LOC101929552 MIR4525 TEPSIN BAHCC1 MRPL12 ASPSCR1 MYADML2 MCRIP1 UTS2R LRRC45 ARL16 ALYREF LOC105371944 LOC100130370 ANAPC11 FASN FSCN2 MIR4740 SLC25A10 TSPAN10 MIR3186 WDR45B AATK PYCR1 PDE6G MAFG DUS1L NOTUM NPB SECTM1 RAB40B HGS LINC00482 MAFG-AS1 CD7 STRA13 ARHGDIA FN3K CSNK1D FAAP100 C17orf89 TBCD RAC3 |
nsv1067160 | Chr.17:82381853 - 82446450 on Build GRCh38 | Loss | C17orf62 HEXDC OGFOD3 LOC105376790 |
nsv828143 | Chr.17:82400094 - 82429642 on Build GRCh38 | Loss | HEXDC OGFOD3 |
nsv469561 | Chr.17:82251148 - 82472274 on Build GRCh38 | Loss | C17orf62 SECTM1 HEXDC UTS2R OGFOD3 CSNK1D LOC101929511 TEX19 LOC105376790 NARF CD7 |
nsv952394 | Chr.17:82297625 - 82408424 on Build GRCh38 | Deletion | SECTM1 UTS2R OGFOD3 TEX19 LOC105376790 CD7 |
nsv1067194 | Chr.17:82294731 - 82653129 on Build GRCh38 | Gain | C17orf62 SECTM1 HEXDC UTS2R OGFOD3 TEX19 WDR45B FOXK2 LOC105376790 NARF CD7 |
esv2762577 | Chr.17:82244836 - 82838431 on Build GRCh38 | Gain+Loss | LOC105376791 SECTM1 RAB40B UTS2R ZNF750 FN3KRP FOXK2 NARF CD7 C17orf62 FN3K HEXDC OGFOD3 CSNK1D LOC101929511 TEX19 WDR45B LOC105376790 TBCD LOC101929552 MIR4525 |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | LOC105376791 SLC16A3 P4HB FOXK2 NARF MIR6787 CCDC57 TEX19 SNORD134 PPP1R27 DCXR RFNG ZNF750 FN3KRP C17orf62 PCYT2 HEXDC OGFOD3 LOC101929511 SIRT7 LOC105376790 GPS1 B3GNTL1 LOC101929552 MIR4525 ASPSCR1 MYADML2 MCRIP1 UTS2R LRRC45 ALYREF METRNL LOC105371944 ANAPC11 FASN WDR45B PYCR1 MAFG DUS1L NOTUM NPB SECTM1 RAB40B MAFG-AS1 CD7 STRA13 ARHGDIA FN3K CSNK1D TBCD RAC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |