Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC400627 ACTG1 TEPSIN FSCN2 FAAP100 LOC107985021 SLC38A10 OXLD1 MIR3186 MIR6786 LOC107985022 CCDC137 LINC00482 AATK LOC100130370 LOC100129503 ARL16 MIR657 MIR4740 HGS BAIAP2 LOC101928855 SLC25A10 BAHCC1 LOC105371921 CEP131 MRPL12 C17orf89 MIR3065 AATK-AS1 MIR338 PDE6G LOC105371925 NPLOC4 RPTOR MIR1250 BAIAP2-AS1 TSPAN10 CHMP6 TMEM105 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | AATK-AS1 AATK CEP131 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | DUS1L MAFG GCGR SLC38A10 ZNF750 OXLD1 PPP1R27 C17orf62 AATK WDR45B ARL16 GPS1 NOTUM UTS2R MIR4525 PCYT2 MIR4740 LRRC45 TEX19 HGS LOC101929552 MAFG-AS1 P4HB MYADML2 FOXK2 BAHCC1 CEP131 CSNK1D CD7 TBCD SLC16A3 RAB40B SIRT7 ALYREF MCRIP1 LOC105371925 NPLOC4 ARHGDIA LOC105376790 ACTG1 SECTM1 TEPSIN SNORD134 FSCN2 FAAP100 NPB DCXR MIR3186 RAC3 MIR6786 CCDC137 LINC00482 FN3KRP LOC100130370 B3GNTL1 ASPSCR1 HEXDC PYCR1 RFNG STRA13 OGFOD3 SLC25A10 LOC105376791 MIR6787 FN3K MRPL12 C17orf89 NARF AATK-AS1 FASN CCDC57 PDE6G LOC101929511 LOC105371944 ANAPC11 TSPAN10 TMEM105 |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | AATK-AS1 TEPSIN AATK LOC105371925 SLC38A10 CEP131 C17orf89 |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | MIR3065 AATK-AS1 BAIAP2 AATK MIR338 CEP131 MIR657 MIR1250 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | TEPSIN BAIAP2 SLC38A10 CEP131 C17orf89 LINC00482 MIR3065 AATK-AS1 AATK MIR338 LOC105371925 MIR657 MIR1250 BAIAP2-AS1 TMEM105 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR3065 AATK-AS1 BAIAP2 AATK MIR338 CEP131 MIR657 MIR1250 BAIAP2-AS1 |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | AATK-AS1 TEPSIN LOC105371925 SLC38A10 CEP131 C17orf89 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | ACTG1 MIR4740 TEPSIN FSCN2 FAAP100 BAHCC1 SLC38A10 CEP131 MIR3186 C17orf89 LINC00482 AATK-AS1 AATK LOC100130370 LOC105371925 NPLOC4 TMEM105 |