Assay Details
Target Gene Details
Entrez Gene ID: | 100130520 |
Gene Name: | CD300c molecule-like |
Gene Aliases: |
CD300H, CD300Hs |
Location: |
Chr.17:74557229-74567512 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100130520 | NM_001324073.1 | NP_001311002.1 | ||
NM_001324076.1 | NP_001311005.1 | |||
XM_006722261.3 | XP_006722324.1 | |||
XM_011525569.2 | XP_011523871.1 | |||
LC013475.1 | ||||
LC013476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12e196 | Chr.17:73675043 - 74565539 on Build GRCh38 | Duplication | LOC100130520 CD300A TTYH2 DNAI2 RPL38 MGC16275 LOC100134391 LOC400620 KIF19 CD300LB GPR142 BTBD17 LINC00469 CD300C GPRC5C |
esv2761978 | Chr.17:73835799 - 74684915 on Build GRCh38 | Gain | LOC100130520 CD300A TTYH2 DNAI2 RAB37 RPL38 MGC16275 KIF19 CD300LB GPR142 BTBD17 LOC101928343 C17orf77 CD300C CD300E CD300LD GPRC5C |
esv3641230 | Chr.17:73831787 - 74694861 on Build GRCh38 | Gain | LOC100130520 CD300A TTYH2 DNAI2 RAB37 RPL38 MGC16275 KIF19 CD300LF CD300LB GPR142 BTBD17 LOC101928343 C17orf77 CD300C CD300E CD300LD GPRC5C |
dgv3282n100 | Chr.17:73814343 - 74719597 on Build GRCh38 | Gain | LOC100130520 CD300A TTYH2 DNAI2 RAB37 RPL38 MGC16275 KIF19 CD300LF CD300LB GPR142 BTBD17 LOC101928343 LINC00469 C17orf77 CD300C CD300E CD300LD GPRC5C |
esv33995 | Chr.17:74261053 - 74732093 on Build GRCh38 | Loss | LOC100130520 CD300A TTYH2 DNAI2 RAB37 KIF19 CD300LF CD300LB GPR142 BTBD17 LOC101928343 C17orf77 CD300C CD300E CD300LD GPRC5C |
nsv517569 | Chr.17:73837862 - 74685802 on Build GRCh38 | Gain+Loss | LOC100130520 CD300A TTYH2 DNAI2 RAB37 RPL38 MGC16275 KIF19 CD300LB GPR142 BTBD17 LOC101928343 C17orf77 CD300C CD300E CD300LD GPRC5C |
dgv5768n54 | Chr.17:73826016 - 74706775 on Build GRCh38 | Gain | LOC100130520 CD300A TTYH2 DNAI2 RAB37 RPL38 MGC16275 KIF19 CD300LF CD300LB GPR142 BTBD17 LOC101928343 LINC00469 C17orf77 CD300C CD300E CD300LD GPRC5C |
esv3573820 | Chr.17:73817932 - 74694378 on Build GRCh38 | Gain | LOC100130520 CD300A TTYH2 DNAI2 RAB37 RPL38 MGC16275 KIF19 CD300LF CD300LB GPR142 BTBD17 LOC101928343 LINC00469 C17orf77 CD300C CD300E CD300LD GPRC5C |
esv3641244 | Chr.17:74527395 - 74686883 on Build GRCh38 | Gain | LOC100130520 RAB37 LOC101928343 C17orf77 CD300C CD300E CD300LD CD300LB |
dgv5774n54 | Chr.17:74428381 - 74673021 on Build GRCh38 | Gain | LOC100130520 CD300A RAB37 LOC101928343 C17orf77 CD300C CD300E CD300LD CD300LB GPRC5C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |