Assay Details
Target Gene Details
Entrez Gene ID: | 400618 |
Gene Name: | SOX9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:72071042-72120793 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOX9-AS1 | NR_103737.1 | |||
NR_103738.1 | ||||
AK056229.1 | ||||
AK094963.1 | ||||
AL833139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833530 | Chr.17:72033949 - 72232973 on Build GRCh38 | Gain+Loss | SOX9 LOC102723517 SOX9-AS1 LOC101928205 LINC01152 |
esv3573809 | Chr.17:71977779 - 72157070 on Build GRCh38 | Gain | LOC102723505 SOX9 LOC102723517 SOX9-AS1 LOC101928205 LINC01152 |