Assay Details
Target Gene Details
Entrez Gene ID: | 339229 |
Gene Name: | oxidoreductase like domain containing 1 |
Gene Aliases: |
C17orf90 |
Location: |
Chr.17:81665036-81666637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OXLD1 | NM_001039842.2 | NP_001034931.1 | ||
NM_001304994.1 | 1 | 131 | NP_001291923.1 | |
NM_001304995.1 | 1 | 131 | NP_001291924.1 | |
NM_001304999.1 | 1 | 131 | NP_001291928.1 | |
NR_130936.1 | 1 | 131 | ||
NR_130937.1 | 1 | 131 | ||
NR_130938.1 | 1 | 131 | ||
AY007126.1 | 1 | 129 | ||
BC090923.1 | AAH90923.1 | |||
BE386163.1 | ||||
BU194770.1 | 1 | 89 | ||
DT220680.1 | ||||
HY012937.1 | 1 | 132 | ||
HY056467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | MIR1250 AATK-AS1 CEP131 BAIAP2 LOC100130370 TMEM105 C17orf89 ACTG1 TEPSIN ARL16 LOC105371925 LOC101928855 BAIAP2-AS1 MRPL12 OXLD1 SLC38A10 AATK NPLOC4 HGS LOC107985021 LOC100129503 PDE6G LOC105371921 BAHCC1 MIR3186 CCDC137 FAAP100 LINC00482 MIR6786 TSPAN10 RPTOR CHMP6 SLC25A10 FSCN2 MIR3065 MIR338 LOC107985022 MIR4740 MIR657 LOC400627 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | AATK-AS1 GCGR MIR6787 TMEM105 C17orf89 ACTG1 TEPSIN B3GNTL1 ARL16 DCXR LOC105371925 ZNF750 C17orf62 DUS1L OXLD1 SLC38A10 FN3K AATK NPLOC4 HGS LRRC45 FASN P4HB ANAPC11 FN3KRP PCYT2 BAHCC1 LOC105371944 CCDC137 FOXK2 CD7 LINC00482 MIR6786 LOC101929511 RFNG UTS2R NOTUM MIR4525 OGFOD3 WDR45B SIRT7 TEX19 PPP1R27 CCDC57 CEP131 LOC100130370 LOC105376790 MYADML2 CSNK1D MRPL12 HEXDC SNORD134 SLC16A3 GPS1 ASPSCR1 PDE6G ALYREF NPB NARF MAFG LOC105376791 MIR3186 FAAP100 SECTM1 TSPAN10 PYCR1 MCRIP1 RAB40B SLC25A10 FSCN2 RAC3 ARHGDIA LOC101929552 MAFG-AS1 MIR4740 TBCD STRA13 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | GCGR MYADML2 ARL16 DCXR DUS1L MRPL12 OXLD1 NPLOC4 HGS LRRC45 GPS1 P4HB ANAPC11 ASPSCR1 PDE6G PCYT2 ALYREF NPB MAFG CCDC137 FAAP100 MIR6786 RFNG TSPAN10 NOTUM PYCR1 MCRIP1 SLC25A10 FSCN2 RAC3 ARHGDIA SIRT7 MAFG-AS1 STRA13 PPP1R27 |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | GCGR CCDC137 MIR6786 TSPAN10 ARL16 MCRIP1 MRPL12 SLC25A10 OXLD1 HGS P4HB PDE6G PPP1R27 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |