Assay Details
Target Gene Details
Entrez Gene ID: | 114826 |
Gene Name: | SET and MYND domain containing 4 |
Gene Aliases: |
ZMYND21 |
Location: |
Chr.17:1779535-1829896 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMYD4 | NM_052928.2 | NP_443160.2 | ||
XM_011523632.2 | XP_011521934.1 | |||
XM_011523633.2 | XP_011521935.1 | |||
XM_017024121.1 | XP_016879610.1 | |||
AB067523.1 | BAB67829.1 | |||
AK057769.1 | BAB71564.1 | |||
AK092149.1 | ||||
AK095369.1 | BAC04538.1 | |||
BC035077.2 | AAH35077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9490 | Chr.17:1653428 - 1828993 on Build GRCh38 | Loss | PRPF8 SERPINF1 SMYD4 MIR22HG WDR81 MIR22 SERPINF2 TLCD2 |
esv33335 | Chr.17:1779713 - 2048672 on Build GRCh38 | Loss | OVCA2 SMYD4 LOC105371485 RTN4RL1 RPA1 DPH1 |
nsv1065585 | Chr.17:1733183 - 1918769 on Build GRCh38 | Gain | SERPINF1 SMYD4 RPA1 WDR81 SERPINF2 |
nsv833335 | Chr.17:1713955 - 1827844 on Build GRCh38 | Gain+Loss | SERPINF1 SMYD4 MIR22HG WDR81 MIR22 SERPINF2 |
nsv574215 | Chr.17:1779718 - 1824004 on Build GRCh38 | Gain | SMYD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |