Assay Details
Target Gene Details
Entrez Gene ID: | 5345 |
Gene Name: | serpin family F member 2 |
Gene Aliases: |
A2AP, AAP, ALPHA-2-PI, API, PLI |
Location: |
Chr.17:1742808-1755265 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINF2 | NM_000934.3 | NP_000925.2 | ||
NM_001165920.1 | NP_001159392.1 | |||
NM_001165921.1 | NP_001159393.1 | |||
XM_005256701.4 | XP_005256758.2 | |||
XM_017024765.1 | XP_016880254.1 | |||
AK303763.1 | ||||
BC031592.1 | AAH31592.1 | |||
D00116.1 | BAA00070.1 | |||
D00174.1 | BAA00124.1 | |||
J02654.1 | AAA35543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9490 | Chr.17:1653428 - 1828993 on Build GRCh38 | Loss | WDR81 PRPF8 MIR22HG SMYD4 SERPINF1 SERPINF2 TLCD2 MIR22 |
nsv1065585 | Chr.17:1733183 - 1918769 on Build GRCh38 | Gain | RPA1 WDR81 SMYD4 SERPINF1 SERPINF2 |
nsv1946 | Chr.17:1717490 - 1781570 on Build GRCh38 | Insertion | WDR81 SMYD4 SERPINF1 SERPINF2 |
nsv516886 | Chr.17:1708905 - 1754359 on Build GRCh38 | Loss | WDR81 MIR22HG SERPINF2 TLCD2 MIR22 |
nsv833335 | Chr.17:1713955 - 1827844 on Build GRCh38 | Gain+Loss | WDR81 MIR22HG SMYD4 SERPINF1 SERPINF2 MIR22 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |