Assay Details
Target Gene Details
Entrez Gene ID: | 9625 |
Gene Name: | apoptosis associated tyrosine kinase |
Gene Aliases: |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
Location: |
Chr.17:81117295-81166072 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK | NM_001080395.2 | NP_001073864.2 | ||
NM_004920.2 | NP_004911.2 | |||
XM_006722193.2 | XP_006722256.1 | |||
XM_006722194.2 | XP_006722257.1 | |||
XM_006722195.2 | XP_006722258.1 | |||
XM_006722196.2 | XP_006722259.1 | |||
XM_011525509.2 | XP_011523811.1 | |||
XM_011525510.2 | XP_011523812.1 | |||
XM_011525511.2 | XP_011523813.1 | |||
XM_017025421.1 | XP_016880910.1 | |||
XM_017025422.1 | XP_016880911.1 | |||
AB014541.2 | ||||
AK094931.1 | ||||
AK131395.1 | BAD18544.1 | |||
AK131529.1 | BAD18667.1 | |||
BC047378.1 | AAH47378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2716426 | Chr.17:81119453 - 81119928 on Build GRCh38 | Deletion |
![]() |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv523417 | Chr.17:81115790 - 81164271 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)