Assay Details
Target Gene Details
Entrez Gene ID: | 79058 |
Gene Name: | ASPSCR1, UBX domain containing tether for SLC2A4 |
Gene Aliases: |
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9 |
Location: |
Chr.17:81977550-82018929 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASPSCR1 | NM_001251888.1 | NP_001238817.1 | ||
NM_024083.3 | NP_076988.1 | |||
NR_045351.1 | ||||
XM_011523601.2 | XP_011521903.1 | |||
XM_011523602.2 | XP_011521904.1 | |||
XM_017025036.1 | XP_016880525.1 | |||
XM_017025037.1 | XP_016880526.1 | |||
XM_017025038.1 | XP_016880527.1 | |||
XM_017025039.1 | XP_016880528.1 | |||
XM_017025040.1 | XP_016880529.1 | |||
XM_017025041.1 | XP_016880530.1 | |||
XM_017025042.1 | XP_016880531.1 | |||
XM_017025043.1 | XP_016880532.1 | |||
XM_017025044.1 | XP_016880533.1 | |||
AF324219.2 | AAK08959.2 | |||
AK057403.1 | BAB71472.1 | |||
AK057851.1 | BAB71595.1 | |||
AK226086.1 | ||||
AK290624.1 | ||||
BC006152.1 | AAH06152.1 | |||
BC018722.1 | AAH18722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952392 | Chr.17:81885825 - 82065724 on Build GRCh38 | Deletion | ANAPC11 ALYREF MAFG GPS1 DCXR STRA13 NPB RAC3 NOTUM RFNG LRRC45 DUS1L SIRT7 MAFG-AS1 MYADML2 PYCR1 ASPSCR1 PCYT2 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MIR4525 OGFOD3 CEP131 ALYREF FASN LOC105371944 FOXK2 SECTM1 C17orf62 DCXR NOTUM TEPSIN LOC105376790 SLC16A3 CCDC137 MIR4740 HEXDC MYADML2 RAB40B MCRIP1 PCYT2 TEX19 AATK ACTG1 HGS FAAP100 GPS1 SNORD134 LOC100130370 MRPL12 NARF TMEM105 MIR6787 BAHCC1 SIRT7 LOC101929552 TBCD WDR45B ANAPC11 MAFG FSCN2 CCDC57 LINC00482 C17orf89 FN3KRP SLC38A10 RAC3 AATK-AS1 OXLD1 NPLOC4 RFNG LOC105376791 LOC105371925 DUS1L CSNK1D MAFG-AS1 MIR6786 PYCR1 ASPSCR1 MIR3186 FN3K LOC101929511 PPP1R27 ARL16 P4HB ARHGDIA GCGR STRA13 PDE6G ZNF750 B3GNTL1 NPB TSPAN10 SLC25A10 LRRC45 CD7 UTS2R |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | ANAPC11 ALYREF MAFG FSCN2 DCXR RAC3 OXLD1 NPLOC4 NOTUM RFNG CCDC137 DUS1L MAFG-AS1 MYADML2 MIR6786 PYCR1 ASPSCR1 MCRIP1 PCYT2 PPP1R27 ARL16 HGS P4HB ARHGDIA FAAP100 GCGR GPS1 STRA13 MRPL12 PDE6G NPB TSPAN10 SLC25A10 LRRC45 SIRT7 |
nsv833569 | Chr.17:81838937 - 82011412 on Build GRCh38 | Loss | ANAPC11 ALYREF MAFG P4HB ARHGDIA NPB NOTUM SIRT7 MAFG-AS1 MYADML2 PYCR1 ASPSCR1 PCYT2 |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | MIR4525 OGFOD3 ALYREF FASN LOC105371944 FOXK2 SECTM1 C17orf62 DCXR NOTUM LOC105376790 SLC16A3 HEXDC MYADML2 RAB40B MCRIP1 PCYT2 TEX19 GPS1 SNORD134 NARF MIR6787 SIRT7 LOC101929552 TBCD WDR45B ANAPC11 METRNL MAFG CCDC57 FN3KRP RAC3 RFNG LOC105376791 DUS1L CSNK1D MAFG-AS1 PYCR1 ASPSCR1 FN3K LOC101929511 PPP1R27 P4HB ARHGDIA STRA13 ZNF750 B3GNTL1 NPB LRRC45 CD7 UTS2R |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |