Assay Details
Target Gene Details
Entrez Gene ID: | 339229 |
Gene Name: | oxidoreductase like domain containing 1 |
Gene Aliases: |
C17orf90 |
Location: |
Chr.17:81665036-81666637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OXLD1 | NM_001039842.2 | NP_001034931.1 | ||
NM_001304994.1 | NP_001291923.1 | |||
NM_001304995.1 | NP_001291924.1 | |||
NM_001304999.1 | 1 | 503 | NP_001291928.1 | |
NR_130936.1 | ||||
NR_130937.1 | ||||
NR_130938.1 | ||||
NR_130939.1 | ||||
AY007126.1 | 1 | 501 | ||
BC090923.1 | AAH90923.1 | |||
BE386163.1 | ||||
BU194770.1 | ||||
CB048026.1 | ||||
DT220680.1 | ||||
HY012937.1 | ||||
HY056467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LINC00482 PDE6G LOC105371921 CHMP6 SLC25A10 AATK-AS1 SLC38A10 ARL16 MIR6786 TMEM105 NPLOC4 LOC100130370 C17orf89 MIR657 CEP131 LOC101928855 MIR1250 ACTG1 HGS BAHCC1 MRPL12 TSPAN10 MIR338 BAIAP2-AS1 MIR4740 AATK TEPSIN MIR3186 OXLD1 FAAP100 FSCN2 MIR3065 LOC105371925 LOC400627 RPTOR LOC100129503 CCDC137 LOC107985021 BAIAP2 LOC107985022 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | RAC3 FN3KRP FOXK2 SLC38A10 ARL16 TMEM105 PCYT2 LOC100130370 C17orf89 TBCD MAFG LOC101929552 FN3K HEXDC CEP131 LOC101929511 ACTG1 GCGR MRPL12 TEX19 NARF NOTUM ZNF750 AATK TEPSIN MIR3186 GPS1 FAAP100 STRA13 CCDC57 CD7 PYCR1 MCRIP1 CCDC137 SLC16A3 CSNK1D MIR6787 MYADML2 LOC105371944 PPP1R27 SNORD134 ARHGDIA LINC00482 PDE6G SLC25A10 RFNG AATK-AS1 MIR6786 NPLOC4 LOC105376790 ANAPC11 ALYREF B3GNTL1 NPB OGFOD3 SIRT7 DCXR LRRC45 WDR45B RAB40B SECTM1 MAFG-AS1 HGS ASPSCR1 BAHCC1 MIR4525 DUS1L TSPAN10 MIR4740 C17orf62 OXLD1 P4HB FSCN2 UTS2R FASN LOC105371925 LOC105376791 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | RAC3 PDE6G SLC25A10 RFNG ARL16 MIR6786 NPLOC4 PCYT2 MAFG ANAPC11 ALYREF NPB SIRT7 DCXR LRRC45 MAFG-AS1 HGS GCGR ASPSCR1 DUS1L MRPL12 TSPAN10 NOTUM GPS1 OXLD1 P4HB FAAP100 FSCN2 STRA13 PYCR1 MCRIP1 CCDC137 MYADML2 PPP1R27 ARHGDIA |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | HGS PDE6G SLC25A10 GCGR ARL16 MRPL12 MIR6786 TSPAN10 OXLD1 P4HB MCRIP1 CCDC137 PPP1R27 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |