Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC101928855 HGS BAIAP2 MIR3065 MIR338 LOC107985021 SLC25A10 LOC400627 AATK MIR657 ACTG1 CCDC137 LINC00482 CEP131 PDE6G FSCN2 CHMP6 TMEM105 MIR3186 TEPSIN BAIAP2-AS1 MIR4740 OXLD1 FAAP100 LOC100130370 ARL16 LOC105371925 MIR1250 SLC38A10 MIR6786 NPLOC4 MRPL12 BAHCC1 LOC100129503 LOC105371921 RPTOR AATK-AS1 C17orf89 LOC107985022 TSPAN10 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | AATK AATK-AS1 CEP131 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | RFNG NPB UTS2R MCRIP1 LOC105376791 PYCR1 RAC3 LOC101929552 MIR4525 LINC00482 CEP131 PDE6G CSNK1D FSCN2 PPP1R27 MIR3186 NOTUM B3GNTL1 OXLD1 ARL16 SLC16A3 LRRC45 MIR6786 NPLOC4 BAHCC1 STRA13 AATK-AS1 C17orf89 HEXDC OGFOD3 C17orf62 FN3KRP P4HB ASPSCR1 HGS TEX19 LOC105371944 SNORD134 MAFG-AS1 SLC25A10 ANAPC11 MAFG GCGR AATK ARHGDIA WDR45B RAB40B TBCD PCYT2 SIRT7 ACTG1 ZNF750 CCDC137 DUS1L FASN ALYREF CD7 TMEM105 FOXK2 LOC105376790 TEPSIN CCDC57 NARF MIR4740 FAAP100 MYADML2 LOC100130370 LOC105371925 SLC38A10 FN3K MRPL12 LOC101929511 SECTM1 GPS1 DCXR MIR6787 TSPAN10 |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | AATK TEPSIN AATK-AS1 C17orf89 LOC105371925 SLC38A10 CEP131 |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | AATK MIR657 AATK-AS1 BAIAP2 MIR3065 MIR338 MIR1250 CEP131 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | TMEM105 TEPSIN BAIAP2-AS1 BAIAP2 MIR3065 MIR338 LOC105371925 MIR1250 SLC38A10 AATK MIR657 AATK-AS1 C17orf89 LINC00482 CEP131 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | AATK BAIAP2-AS1 MIR657 AATK-AS1 BAIAP2 MIR3065 MIR338 MIR1250 CEP131 |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | TEPSIN AATK-AS1 C17orf89 LOC105371925 SLC38A10 CEP131 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | TMEM105 MIR3186 TEPSIN MIR4740 FAAP100 LOC100130370 LOC105371925 SLC38A10 NPLOC4 BAHCC1 AATK AATK-AS1 C17orf89 ACTG1 LINC00482 CEP131 FSCN2 |