Assay Details
Target Gene Details
Entrez Gene ID: | 10458 |
Gene Name: | BAI1 associated protein 2 |
Gene Aliases: |
BAP2, FLAF3, IRSP53 |
Location: |
Chr.17:81035147-81117432 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | MIR1250 AATK LOC107985021 SLC38A10 LOC100129503 FAAP100 LOC105371921 MRPL12 OXLD1 MIR6786 TSPAN10 MIR338 CCDC137 ACTG1 C17orf89 SLC25A10 MIR3186 TMEM105 LOC105371925 CEP131 MIR3065 AATK-AS1 MIR4740 LOC107985022 LOC100130370 NPLOC4 TEPSIN PDE6G BAHCC1 RPTOR CHMP6 LOC400627 HGS LOC101928855 ARL16 BAIAP2-AS1 LINC00482 BAIAP2 MIR657 FSCN2 |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | MIR1250 AATK BAIAP2-AS1 BAIAP2 MIR338 MIR3065 MIR657 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | MIR1250 AATK SLC38A10 TEPSIN MIR338 BAIAP2-AS1 LINC00482 C17orf89 TMEM105 BAIAP2 LOC105371925 CEP131 MIR3065 MIR657 AATK-AS1 |
nsv576169 | Chr.17:81030816 - 81043967 on Build GRCh38 | Loss | BAIAP2-AS1 BAIAP2 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR1250 AATK BAIAP2-AS1 BAIAP2 CEP131 MIR338 MIR3065 MIR657 AATK-AS1 |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | MIR1250 AATK BAIAP2-AS1 BAIAP2 MIR338 MIR3065 MIR657 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |