Assay Details
Target Gene Details
Entrez Gene ID: | 100131439 |
Gene Name: | CD300 molecule like family member d |
Gene Aliases: |
CD300D, CLM-4, CMRF35-A4, CMRF35A4 |
Location: |
Chr.17:74579972-74592231 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD300LD | NM_001115152.1 | NP_001108624.1 | ||
AY358148.1 | AAQ88515.1 | |||
EF137868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761978 | Chr.17:73835799 - 74684915 on Build GRCh38 | Gain | GPR142 GPRC5C BTBD17 KIF19 RPL38 CD300LD DNAI2 MGC16275 CD300LB LOC100130520 TTYH2 CD300E LOC101928343 C17orf77 RAB37 CD300A CD300C |
esv3641230 | Chr.17:73831787 - 74694861 on Build GRCh38 | Gain | GPR142 GPRC5C BTBD17 KIF19 RPL38 CD300LD DNAI2 MGC16275 CD300LF CD300LB LOC100130520 TTYH2 CD300E LOC101928343 C17orf77 RAB37 CD300A CD300C |
dgv3282n100 | Chr.17:73814343 - 74719597 on Build GRCh38 | Gain | GPR142 GPRC5C BTBD17 KIF19 RPL38 CD300LD DNAI2 MGC16275 CD300LF CD300LB LOC100130520 TTYH2 CD300E LINC00469 LOC101928343 C17orf77 RAB37 CD300A CD300C |
esv33995 | Chr.17:74261053 - 74732093 on Build GRCh38 | Loss | GPR142 GPRC5C BTBD17 KIF19 CD300LD DNAI2 CD300LF CD300LB LOC100130520 TTYH2 CD300E LOC101928343 C17orf77 RAB37 CD300A CD300C |
nsv517569 | Chr.17:73837862 - 74685802 on Build GRCh38 | Gain+Loss | GPR142 GPRC5C BTBD17 KIF19 RPL38 CD300LD DNAI2 MGC16275 CD300LB LOC100130520 TTYH2 CD300E LOC101928343 C17orf77 RAB37 CD300A CD300C |
dgv5768n54 | Chr.17:73826016 - 74706775 on Build GRCh38 | Gain | GPR142 GPRC5C BTBD17 KIF19 RPL38 CD300LD DNAI2 MGC16275 CD300LF CD300LB LOC100130520 TTYH2 CD300E LINC00469 LOC101928343 C17orf77 RAB37 CD300A CD300C |
esv3573820 | Chr.17:73817932 - 74694378 on Build GRCh38 | Gain | GPR142 GPRC5C BTBD17 KIF19 RPL38 CD300LD DNAI2 MGC16275 CD300LF CD300LB LOC100130520 TTYH2 CD300E LINC00469 LOC101928343 C17orf77 RAB37 CD300A CD300C |
esv3641244 | Chr.17:74527395 - 74686883 on Build GRCh38 | Gain | CD300E LOC101928343 CD300LD C17orf77 RAB37 CD300LB CD300C LOC100130520 |
dgv5774n54 | Chr.17:74428381 - 74673021 on Build GRCh38 | Gain | CD300E GPRC5C LOC101928343 CD300LD C17orf77 RAB37 CD300A CD300LB CD300C LOC100130520 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |