Assay Details
Target Gene Details
Entrez Gene ID: | 5148 |
Gene Name: | phosphodiesterase 6G |
Gene Aliases: |
PDEG, RP57 |
Location: |
Chr.17:81650459-81663418 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE6G | NM_002602.3 | NP_002593.1 | ||
NR_026872.1 | ||||
XM_006721942.3 | XP_006722005.1 | |||
XM_017024734.1 | XP_016880223.1 | |||
BC106884.2 | AAI06885.1 | |||
BQ638010.1 | ||||
BQ639132.1 | ||||
BX537414.1 | CAD97656.1 | |||
BX647779.1 | ||||
M36476.1 | AAA60044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | TMEM105 MIR3186 MIR657 MIR338 MIR4740 BAHCC1 CCDC137 TEPSIN SLC38A10 BAIAP2 BAIAP2-AS1 LOC105371921 ARL16 OXLD1 FSCN2 CEP131 LOC107985022 MRPL12 LINC00482 C17orf89 AATK ACTG1 SLC25A10 LOC100129503 MIR1250 RPTOR LOC101928855 MIR3065 LOC100130370 LOC107985021 MIR6786 CHMP6 PDE6G LOC400627 HGS LOC105371925 TSPAN10 FAAP100 AATK-AS1 NPLOC4 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MIR3186 RAC3 CSNK1D MIR4740 NOTUM CCDC137 PPP1R27 OGFOD3 MCRIP1 TEX19 ARL16 OXLD1 MAFG-AS1 LOC101929552 CEP131 RAB40B ALYREF PCYT2 SECTM1 C17orf89 LOC101929511 AATK P4HB LOC105376791 SLC25A10 CD7 MIR6787 PDE6G WDR45B HGS LOC105371925 TSPAN10 FAAP100 DCXR FOXK2 SIRT7 RFNG TBCD UTS2R NPLOC4 TMEM105 MYADML2 BAHCC1 LOC105376790 ARHGDIA STRA13 TEPSIN SLC38A10 FASN GPS1 NARF FSCN2 NPB MRPL12 LINC00482 ASPSCR1 SNORD134 ACTG1 FN3K MIR4525 DUS1L PYCR1 GCGR LOC100130370 ANAPC11 LRRC45 MIR6786 C17orf62 SLC16A3 CCDC57 LOC105371944 ZNF750 FN3KRP HEXDC AATK-AS1 B3GNTL1 MAFG |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | MYADML2 RAC3 ARHGDIA NOTUM STRA13 CCDC137 PPP1R27 MCRIP1 ARL16 OXLD1 MAFG-AS1 GPS1 FSCN2 NPB ALYREF PCYT2 MRPL12 ASPSCR1 P4HB SLC25A10 DUS1L PYCR1 GCGR ANAPC11 LRRC45 MIR6786 PDE6G HGS TSPAN10 FAAP100 DCXR SIRT7 RFNG NPLOC4 MAFG |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | SLC25A10 GCGR CCDC137 PPP1R27 MCRIP1 MIR6786 ARL16 PDE6G OXLD1 HGS TSPAN10 MRPL12 P4HB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |