Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | NPLOC4 MIR4740 LOC105371925 SLC25A10 PDE6G CHMP6 TMEM105 FSCN2 AATK SLC38A10 AATK-AS1 MIR1250 MIR3065 BAIAP2-AS1 TSPAN10 C17orf89 CCDC137 ARL16 ACTG1 MIR338 LOC101928855 FAAP100 LINC00482 OXLD1 RPTOR HGS LOC100130370 LOC100129503 LOC105371921 TEPSIN MIR3186 MRPL12 BAHCC1 MIR6786 LOC400627 CEP131 MIR657 LOC107985021 BAIAP2 LOC107985022 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | CEP131 AATK AATK-AS1 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | FOXK2 NPLOC4 TEX19 LOC105371925 SLC25A10 PDE6G MIR4525 TMEM105 AATK SLC38A10 AATK-AS1 ZNF750 DUS1L PPP1R27 WDR45B PCYT2 CSNK1D NARF RAC3 FAAP100 LINC00482 B3GNTL1 TBCD RFNG MCRIP1 LOC105371944 HGS MIR6787 SNORD134 MIR3186 MRPL12 C17orf62 BAHCC1 FN3K LOC101929552 CCDC57 CEP131 DCXR STRA13 CD7 MIR4740 OGFOD3 ASPSCR1 FSCN2 GPS1 SECTM1 TSPAN10 C17orf89 LOC105376791 CCDC137 ARL16 SLC16A3 PYCR1 ACTG1 UTS2R MYADML2 FN3KRP P4HB OXLD1 LOC101929511 LOC100130370 ANAPC11 GCGR MAFG TEPSIN SIRT7 MIR6786 ARHGDIA NPB LRRC45 RAB40B HEXDC NOTUM LOC105376790 MAFG-AS1 FASN ALYREF |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | LOC105371925 CEP131 AATK TEPSIN SLC38A10 AATK-AS1 C17orf89 |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | CEP131 MIR657 MIR338 AATK AATK-AS1 MIR1250 MIR3065 BAIAP2 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | LOC105371925 TMEM105 AATK TEPSIN SLC38A10 AATK-AS1 MIR1250 MIR3065 BAIAP2-AS1 C17orf89 CEP131 MIR657 MIR338 LINC00482 BAIAP2 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | CEP131 MIR657 MIR338 AATK AATK-AS1 MIR1250 MIR3065 BAIAP2 BAIAP2-AS1 |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | LOC105371925 CEP131 TEPSIN SLC38A10 AATK-AS1 C17orf89 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | NPLOC4 MIR4740 LOC105371925 LOC100130370 TMEM105 FSCN2 AATK TEPSIN SLC38A10 AATK-AS1 MIR3186 C17orf89 BAHCC1 CEP131 ACTG1 FAAP100 LINC00482 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | MIR657 MIR338 AATK AATK-AS1 MIR1250 MIR3065 BAIAP2 |