Assay Details
Target Gene Details
Entrez Gene ID: | 9625 |
Gene Name: | apoptosis associated tyrosine kinase |
Gene Aliases: |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
Location: |
Chr.17:81117295-81166072 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK | NM_001080395.2 | NP_001073864.2 | ||
NM_004920.2 | NP_004911.2 | |||
XM_006722193.2 | XP_006722256.1 | |||
XM_006722194.2 | XP_006722257.1 | |||
XM_006722195.2 | XP_006722258.1 | |||
XM_006722196.2 | XP_006722259.1 | |||
XM_011525509.2 | XP_011523811.1 | |||
XM_011525510.2 | XP_011523812.1 | |||
XM_011525511.2 | XP_011523813.1 | |||
XM_017025421.1 | XP_016880910.1 | |||
XM_017025422.1 | XP_016880911.1 | |||
AB014541.2 | ||||
AK094931.1 | ||||
AK131395.1 | BAD18544.1 | |||
AK131529.1 | BAD18667.1 | |||
BC047378.1 | AAH47378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | RPTOR LOC107985022 TMEM105 PDE6G AATK-AS1 MIR6786 NPLOC4 LOC105371921 LOC105371925 MIR1250 AATK MIR3065 BAIAP2 TSPAN10 CCDC137 MIR657 SLC25A10 CHMP6 ARL16 FSCN2 BAHCC1 OXLD1 LOC400627 MRPL12 LOC107985021 FAAP100 MIR338 SLC38A10 CEP131 HGS LOC100130370 MIR4740 LINC00482 MIR3186 LOC101928855 ACTG1 LOC100129503 TEPSIN C17orf89 BAIAP2-AS1 |
esv2716425 | Chr.17:81119070 - 81119204 on Build GRCh38 | Deletion | AATK |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | MIR1250 AATK MIR3065 BAIAP2 AATK-AS1 MIR338 CEP131 MIR657 |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | MIR1250 AATK MIR3065 BAIAP2 MIR338 MIR657 BAIAP2-AS1 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | TMEM105 AATK-AS1 MIR338 SLC38A10 CEP131 LINC00482 LOC105371925 MIR1250 AATK MIR3065 BAIAP2 TEPSIN C17orf89 MIR657 BAIAP2-AS1 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR1250 AATK MIR3065 BAIAP2 AATK-AS1 MIR338 CEP131 MIR657 BAIAP2-AS1 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | MIR1250 AATK MIR3065 BAIAP2 AATK-AS1 MIR338 MIR657 |
nsv523417 | Chr.17:81115790 - 81164271 on Build GRCh38 | Gain | MIR1250 AATK MIR3065 BAIAP2 MIR338 MIR657 |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | MIR1250 AATK MIR3065 BAIAP2 MIR338 MIR657 BAIAP2-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |