Assay Details
Target Gene Details
Entrez Gene ID: | 79058 |
Gene Name: | ASPSCR1, UBX domain containing tether for SLC2A4 |
Gene Aliases: |
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9 |
Location: |
Chr.17:81977550-82018929 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASPSCR1 | NM_001251888.1 | NP_001238817.1 | ||
NM_024083.3 | NP_076988.1 | |||
NR_045351.1 | ||||
XM_011523601.2 | XP_011521903.1 | |||
XM_011523602.2 | XP_011521904.1 | |||
XM_017025036.1 | XP_016880525.1 | |||
XM_017025037.1 | XP_016880526.1 | |||
XM_017025038.1 | XP_016880527.1 | |||
XM_017025039.1 | XP_016880528.1 | |||
XM_017025040.1 | XP_016880529.1 | |||
XM_017025041.1 | XP_016880530.1 | |||
XM_017025042.1 | XP_016880531.1 | |||
XM_017025043.1 | XP_016880532.1 | |||
XM_017025044.1 | XP_016880533.1 | |||
AF324219.2 | AAK08959.2 | |||
AK057403.1 | BAB71472.1 | |||
AK057851.1 | BAB71595.1 | |||
AK226086.1 | ||||
AK290624.1 | ||||
BC006152.1 | AAH06152.1 | |||
BC018722.1 | AAH18722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952392 | Chr.17:81885825 - 82065724 on Build GRCh38 | Deletion | MAFG GPS1 RFNG PYCR1 ASPSCR1 NPB MYADML2 ALYREF PCYT2 MAFG-AS1 NOTUM DCXR LRRC45 SIRT7 ANAPC11 DUS1L RAC3 STRA13 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | RFNG TEX19 CCDC137 FASN NPLOC4 SNORD134 LOC105376790 MIR6787 LOC101929552 NOTUM DCXR LRRC45 SIRT7 DUS1L CCDC57 LOC101929511 OXLD1 PYCR1 TBCD ACTG1 NPB SECTM1 FAAP100 MIR4740 ARL16 MAFG-AS1 TMEM105 LOC105371925 B3GNTL1 AATK-AS1 NARF WDR45B MCRIP1 CEP131 TSPAN10 GCGR C17orf62 MIR6786 HEXDC MAFG GPS1 CD7 SLC25A10 TEPSIN SLC38A10 ALYREF PCYT2 HGS RAB40B PPP1R27 UTS2R LOC100130370 ZNF750 AATK FOXK2 MRPL12 OGFOD3 SLC16A3 P4HB LOC105376791 ASPSCR1 BAHCC1 CSNK1D LINC00482 MYADML2 FSCN2 PDE6G LOC105371944 MIR3186 FN3KRP ANAPC11 ARHGDIA MIR4525 RAC3 FN3K STRA13 C17orf89 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | MAFG GPS1 RFNG CCDC137 SLC25A10 ALYREF PCYT2 HGS PPP1R27 NPLOC4 NOTUM MRPL12 DCXR LRRC45 SIRT7 DUS1L P4HB OXLD1 PYCR1 ASPSCR1 NPB MYADML2 FAAP100 FSCN2 PDE6G ARL16 MAFG-AS1 MCRIP1 ANAPC11 ARHGDIA TSPAN10 GCGR RAC3 MIR6786 STRA13 |
nsv833569 | Chr.17:81838937 - 82011412 on Build GRCh38 | Loss | MAFG PYCR1 ASPSCR1 NPB MYADML2 ALYREF PCYT2 MAFG-AS1 NOTUM SIRT7 ANAPC11 ARHGDIA P4HB |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | RFNG TEX19 FASN SNORD134 LOC105376790 MIR6787 LOC101929552 NOTUM DCXR LRRC45 SIRT7 DUS1L CCDC57 LOC101929511 PYCR1 TBCD NPB SECTM1 MAFG-AS1 B3GNTL1 NARF WDR45B MCRIP1 C17orf62 HEXDC MAFG GPS1 CD7 ALYREF PCYT2 RAB40B PPP1R27 UTS2R ZNF750 FOXK2 OGFOD3 SLC16A3 P4HB LOC105376791 ASPSCR1 CSNK1D METRNL MYADML2 LOC105371944 FN3KRP ANAPC11 ARHGDIA MIR4525 RAC3 FN3K STRA13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |