Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3346567 | Chr.16:458004 - 458327 on Build GRCh38 | Duplication | RAB11FIP3 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR6768 HBA1 FBXL16 RAB26 RPS2 HBZ PRSS27 PRR35 LOC107984876 FLJ42627 CASKIN1 LOC100287175 RNPS1 ZNF598 FAM234A IFT140 PKD1 SLC9A3R2 GFER FAHD1 HAGHL LOC105371184 LMF1 MIR5587 CAPN15 BRICD5 TELO2 MIR3677 UBE2I LOC652276 TPSG1 DNASE1L2 MIR6767 HN1L RAB40C HBQ1 LOC105371046 SNORA78 PIGQ NARFL TPSB2 MRPL28 MLST8 ATP6V0C NDUFB10 MIR4717 METRN TMEM8A AXIN1 RGS11 MIR3176 ARHGDIG E4F1 TPSAB1 LOC106660606 MIR4516 RPUSD1 UNKL PRSS29P CACNA1H HS3ST6 C1QTNF8 TMEM204 BAIAP3 PTX4 LOC100134368 C16orf91 MIR3180-5 LINC00235 RPL3L KCTD5 ABCA3 MSLN FAM173A TPSD1 MPG TSR3 LOC101929280 SSTR5 PGP MIR3177 SNHG19 NHLRC4 RHBDL1 TSC2 MCRIP2 HAGH EME2 SNORD60 MEIOB MIR6511B1 WDR24 CRAMP1 NPRL3 HBM SNORA10 LUC7L GNG13 C16orf59 STUB1 DDX11L10 WDR90 METTL26 LOC105371045 SPSB3 NPW TBL3 SOX8 MIR6859-4 MIR662 PDIA2 CHTF18 NTN3 NME3 CCNF PDPK1 SYNGR3 JMJD8 WFIKKN1 RHOT2 CCDC78 GNPTG MIR3178 MAPK8IP3 MIR940 RAB11FIP3 SNHG9 NME4 SSTR5-AS1 LOC729652 LOC105371047 MSRB1 CLCN7 LOC107987233 TRAF7 LMF1-AS1 ERVK13-1 POLR3K MIR1225 NTHL1 SNRNP25 MRPS34 NUBP2 PRR25 IGFALS RPL23AP5 LINC00254 ECI1 SNORA64 CEMP1 TBC1D24 HBA2 RHBDF1 AMDHD2 LOC105371049 WASIR2 CCDC154 LOC105371038 RNF151 NOXO1 ABCA17P DECR2 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | TMEM8A AXIN1 RGS11 ARHGDIG MRPL28 RAB11FIP3 RPL23AP5 NME4 LOC100134368 FAM234A PDIA2 DECR2 |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | RAB40C PIGQ NARFL FBXL16 NHLRC4 RAB11FIP3 RHBDL1 MCRIP2 PRR35 WDR24 METRN MIR3176 LOC107987233 LOC100287175 RPUSD1 GNG13 STUB1 WDR90 METTL26 PRR25 HAGHL LINC00235 MIR662 CHTF18 LOC105371184 MIR5587 MSLN FAM173A JMJD8 CAPN15 WFIKKN1 LOC105371038 RHOT2 LOC101929280 CCDC78 |
nsv1700 | Chr.16:457843 - 476640 on Build GRCh38 | Insertion | RAB11FIP3 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | TMEM8A AXIN1 MRPL28 RAB11FIP3 CAPN15 RPL23AP5 NME4 LOC100134368 LINC00235 DECR2 |
esv1003529 | Chr.16:455611 - 459074 on Build GRCh38 | Insertion | RAB11FIP3 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | TMEM8A AXIN1 RGS11 ARHGDIG MRPL28 RAB11FIP3 RPL23AP5 NME4 LOC100134368 FAM234A PDIA2 DECR2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |