Assay Details
Target Gene Details
Entrez Gene ID: | 81631 |
Gene Name: | microtubule associated protein 1 light chain 3 beta |
Gene Aliases: |
ATG8F, LC3B, MAP1A/1BLC3, MAP1LC3B-a |
Location: |
Chr.16:87392195-87404774 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAP1LC3B | NM_022818.4 | NP_073729.1 | ||
AF087871.1 | AAM10499.1 | |||
AF183417.1 | AAG09686.1 | |||
AF303888.1 | AAG23182.1 | |||
AK025556.1 | BAB15169.1 | |||
AK290956.1 | ||||
AK310224.1 | ||||
AK310335.1 | ||||
AL834298.1 | CAD38970.1 | |||
BC018634.2 | AAH18634.1 | |||
BC041874.1 | AAH41874.1 | |||
BC045759.1 | AAH45759.1 | |||
BC067797.1 | AAH67797.1 | |||
BG771621.1 | ||||
DA577457.1 | ||||
DB055844.1 | ||||
JN663879.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv499n67 | Chr.16:87353978 - 87409229 on Build GRCh38 | Gain | MAP1LC3B ZCCHC14 FBXO31 |
nsv833320 | Chr.16:87315844 - 87501226 on Build GRCh38 | Gain | LOC101928737 C16orf95 MAP1LC3B ZCCHC14 FBXO31 LOC101928659 |
nsv428330 | Chr.16:87336922 - 87633082 on Build GRCh38 | Gain | JPH3 LOC101928737 MAP1LC3B ZCCHC14 FBXO31 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |