Assay Details
Target Gene Details
Entrez Gene ID: | 22980 |
Gene Name: | transcription factor 25 |
Gene Aliases: |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
Location: |
Chr.16:89873583-89911384 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573744 | Chr.16:89738447 - 89915950 on Build GRCh38 | Loss | LOC105371419 ZNF276 SPIRE2 TCF25 FANCA LOC107984817 |
esv2758671 | Chr.16:89804324 - 89901541 on Build GRCh38 | Loss | LOC105371419 SPIRE2 TCF25 FANCA LOC107984817 |
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | LOC105371419 TUBB3 GAS8-AS1 SPATA33 SPIRE2 DPEP1 LOC107984817 VPS9D1 CENPBD1 AFG3L1P GAS8 ZNF276 CDK10 CHMP1A DBNDD1 TCF25 FANCA SPATA2L MC1R VPS9D1-AS1 DEF8 |
nsv573810 | Chr.16:89857498 - 90037251 on Build GRCh38 | Loss | CENPBD1 AFG3L1P TUBB3 GAS8 GAS8-AS1 SPIRE2 DBNDD1 TCF25 MC1R DEF8 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | SPG7 TUBB3 ZNF778 SPATA33 SPIRE2 LOC100287036 DPEP1 VPS9D1 CPNE7 SNORD68 ANKRD11 ZNF276 CDK10 CHMP1A FANCA LOC105371414 SPATA2L VPS9D1-AS1 DEF8 LOC105371419 FAM157C GAS8-AS1 LOC107984817 CENPBD1 AFG3L1P LOC101927817 GAS8 RPL13 PRDM7 DBNDD1 TCF25 URAHP MC1R TUBB8P7 |
nsv573799 | Chr.16:89852186 - 90022379 on Build GRCh38 | Gain | CENPBD1 AFG3L1P LOC105371419 TUBB3 GAS8 SPIRE2 DBNDD1 TCF25 MC1R DEF8 |
nsv1066499 | Chr.16:89891957 - 90028891 on Build GRCh38 | Loss | CENPBD1 AFG3L1P TUBB3 GAS8 DBNDD1 TCF25 MC1R DEF8 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC107987238 SPG7 TUBB3 ZNF778 SPATA33 VPS9D1 MVD CDH15 CPNE7 SNORD68 VPS9D1-AS1 SNAI3-AS1 LOC339059 LOC105371419 CBFA2T3 LOC100289580 LOC100129697 GAS8-AS1 LOC107984817 CTU2 CENPBD1 APRT GALNS PIEZO1 LINC00304 PRDM7 TCF25 URAHP TUBB8P7 ACSF3 CDT1 SLC22A31 SPIRE2 LOC100287036 DPEP1 ANKRD11 ZNF276 CDK10 LOC105371409 CHMP1A PABPN1L FANCA LOC105371414 SPATA2L DEF8 SNAI3 FAM157C LOC101927793 IL17C AFG3L1P LOC101927817 RNF166 TRAPPC2L GAS8 RPL13 MIR4722 LOC400558 DBNDD1 MC1R CYBA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |