Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521610 | Chr.16:430096 - 445538 on Build GRCh38 | Gain | RAB11FIP3 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | NOXO1 WDR24 CASKIN1 WFIKKN1 MIR3177 CCDC78 MIR3178 AMDHD2 CHTF18 MEIOB PRR35 SNORA64 WASIR2 FAM173A SOX8 MIR6859-4 CCNF SNHG19 METRN MPG METTL26 LINC00235 LMF1-AS1 TMEM8A NPW AXIN1 LOC100287175 C16orf59 TPSG1 LOC100134368 ATP6V0C MCRIP2 HS3ST6 HAGHL MIR6768 LMF1 LOC105371045 TELO2 PDPK1 SYNGR3 RHOT2 PDIA2 SNORD60 GFER RNPS1 NME4 HN1L RPL3L NME3 TBL3 CAPN15 ECI1 LUC7L NTHL1 MAPK8IP3 LOC107987233 MRPS34 TPSAB1 UBE2I RPS2 LOC106660606 LOC105371038 ABCA17P HAGH TSR3 E4F1 RAB11FIP3 CCDC154 RPL23AP5 NDUFB10 CLCN7 LOC729652 RHBDL1 LOC105371047 MIR6511B1 DDX11L10 FAM234A LINC00254 MSLN RAB26 MIR3677 TBC1D24 PRSS29P ZNF598 HBA2 MIR4516 POLR3K MIR5587 BAIAP3 ABCA3 LOC105371184 RNF151 ARHGDIG CRAMP1 SSTR5-AS1 GNPTG MRPL28 SLC9A3R2 BRICD5 CACNA1H TPSD1 NUBP2 FBXL16 EME2 TPSB2 SSTR5 DNASE1L2 MIR6767 MIR1225 FAHD1 MLST8 HBA1 LOC105371049 LOC105371046 PGP KCTD5 MIR4717 SNRNP25 LOC107984876 STUB1 C16orf91 TMEM204 SNORA10 PRSS27 NARFL IGFALS SNHG9 NPRL3 HBQ1 FLJ42627 RGS11 SNORA78 TRAF7 HBM CEMP1 C1QTNF8 MIR3180-5 MIR3176 LOC652276 RHBDF1 RPUSD1 PRR25 LOC101929280 GNG13 IFT140 JMJD8 NTN3 MIR662 UNKL TSC2 WDR90 HBZ DECR2 PIGQ ERVK13-1 MSRB1 RAB40C NHLRC4 SPSB3 MIR940 PKD1 PTX4 |
dgv4810n54 | Chr.16:380011 - 445662 on Build GRCh38 | Loss | RAB11FIP3 DECR2 NME4 RPL23AP5 TMEM8A LOC100134368 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RAB11FIP3 RGS11 DECR2 PDIA2 MRPL28 NME4 RPL23AP5 FAM234A TMEM8A AXIN1 LOC100134368 ARHGDIG |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | RAB11FIP3 DECR2 MRPL28 LINC00235 NME4 RPL23AP5 TMEM8A AXIN1 CAPN15 LOC100134368 |
nsv457303 | Chr.16:383623 - 445662 on Build GRCh38 | Loss | RAB11FIP3 DECR2 NME4 RPL23AP5 LOC100134368 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | RAB11FIP3 DECR2 MRPL28 NME4 RPL23AP5 TMEM8A AXIN1 LOC100134368 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RAB11FIP3 RGS11 DECR2 PDIA2 MRPL28 NME4 RPL23AP5 FAM234A TMEM8A AXIN1 LOC100134368 ARHGDIG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |