Assay Details
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | NP_001157032.1 | ||
NM_152764.2 | NP_689977.2 | |||
BC029829.1 | AAH29829.1 | |||
CD300613.1 | ||||
DB461414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | HAGH MEIOB FAHD1 |
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss | LINC00254 MEIOB FAHD1 HS3ST6 |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | HAGH MEIOB FAHD1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | DDX11L10 PRSS29P CHTF18 LOC105371184 LOC105371046 HAGHL NARFL NME3 SSTR5-AS1 SNORA78 HBM PIGQ WDR90 NDUFB10 DECR2 RHBDL1 ARHGDIG UBE2I SSTR5 PRR35 LOC101929280 SNRNP25 ABCA3 MRPS34 ERVK13-1 SNORA64 PDPK1 FAM234A TSC2 MIR5587 RAB40C MIR940 MIR4516 TPSD1 RNF151 MEIOB IGFALS MIR3180-5 ATP6V0C TPSG1 SYNGR3 TMEM8A HBZ POLR3K MIR3176 LOC100287175 GNPTG LOC107987233 FBXL16 LOC106660606 MIR3177 MIR662 LINC00254 TMEM204 BAIAP3 PGP GNG13 CAPN15 METRN MIR4717 MCRIP2 TSR3 LINC00235 JMJD8 PRR25 FAHD1 HBQ1 LOC105371045 C16orf91 LOC729652 NME4 METTL26 LUC7L MIR3178 RHOT2 CACNA1H AMDHD2 RGS11 C16orf59 TRAF7 MIR3677 FAM173A NHLRC4 MIR6768 MIR6767 NTN3 NPRL3 MAPK8IP3 UNKL SOX8 HBA2 MIR6511B1 PTX4 TBC1D24 SLC9A3R2 RPUSD1 LOC100134368 KCTD5 TELO2 NPW PKD1 LMF1-AS1 BRICD5 LOC105371047 GFER SNHG9 NUBP2 EME2 LOC105371049 LMF1 NTHL1 LOC105371038 MSLN HBA1 SNHG19 WASIR2 ZNF598 MIR1225 HAGH ABCA17P CCDC78 PRSS27 CCDC154 TPSB2 SNORA10 STUB1 RAB26 MIR6859-4 AXIN1 RPL23AP5 FLJ42627 MRPL28 RPL3L WFIKKN1 SNORD60 WDR24 TBL3 RHBDF1 HS3ST6 RPS2 LOC107984876 ECI1 SPSB3 CASKIN1 RNPS1 MLST8 E4F1 CRAMP1 HN1L TPSAB1 CEMP1 PDIA2 CLCN7 MPG NOXO1 DNASE1L2 LOC652276 C1QTNF8 RAB11FIP3 IFT140 CCNF MSRB1 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | NUBP2 CRAMP1 HN1L EME2 MAPK8IP3 LOC105371046 MRPS34 FAHD1 HAGH NME3 MIR3177 SPSB3 TMEM204 TELO2 MEIOB LOC105371047 IFT140 IGFALS |
nsv1046474 | Chr.16:1846666 - 1873690 on Build GRCh38 | Loss | MEIOB |
nsv519366 | Chr.16:1851210 - 1961125 on Build GRCh38 | Loss | RPL3L LINC00254 NDUFB10 MEIOB MSRB1 HS3ST6 |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | NUBP2 HAGH MEIOB FAHD1 IGFALS |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | HAGH LINC00254 MEIOB FAHD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |