Assay Details
Target Gene Details
Entrez Gene ID: | 1003 |
Gene Name: | cadherin 5 |
Gene Aliases: |
7B4, CD144 |
Location: |
Chr.16:66366607-66404786 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH5 | NM_001795.4 | NP_001786.2 | ||
XM_011522801.1 | XP_011521103.1 | |||
AB035304.1 | BAA87418.1 | |||
AB209908.1 | BAD93145.1 | |||
AK300299.1 | ||||
AK300461.1 | ||||
BC096363.3 | AAH96363.1 | |||
BC096364.3 | ||||
BC096365.1 | AAH96365.1 | |||
BC117520.2 | ||||
DC381809.1 | ||||
DC382718.1 | ||||
U84722.1 | AAB41796.1 | |||
X59796.1 | CAA42468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833264 | Chr.16:66298692 - 66471069 on Build GRCh38 | Gain | CDH5 BEAN1 BEAN1-AS1 LINC00920 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |