Assay Details
Target Gene Details
Entrez Gene ID: | 4913 |
Gene Name: | nth-like DNA glycosylase 1 |
Gene Aliases: |
FAP3, NTH1, OCTS3, hNTH1 |
Location: |
Chr.16:2039815-2047872 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NTHL1 | NM_001318193.1 | NP_001305122.1 | ||
NM_001318194.1 | NP_001305123.1 | |||
NM_002528.6 | NP_002519.1 | |||
XM_017023253.1 | XP_016878742.1 | |||
AB001575.1 | BAA19413.1 | |||
BC000391.2 | AAH00391.2 | |||
BC003014.1 | AAH03014.1 | |||
BE900738.1 | ||||
BG468617.1 | ||||
U79718.1 | AAB41534.1 | |||
U81285.1 | AAC51136.1 | |||
Y09687.1 | CAA70865.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR3677 DNASE1L2 PRSS29P GFER WDR90 METRN DECR2 NTN3 CAPN15 HAGH CHTF18 MIR6768 TSC2 TPSD1 STUB1 LUC7L NDUFB10 HBQ1 SNORA78 GNPTG MRPS34 TBL3 LOC729652 MSLN LOC107984876 SNHG19 FAHD1 LMF1-AS1 BRICD5 FLJ42627 MIR4717 MIR3177 METTL26 NUBP2 SSTR5-AS1 LOC105371047 CLCN7 PIGQ MAPK8IP3 NME4 CCDC154 RHBDF1 MIR662 RAB40C CCNF UBE2I CRAMP1 TPSG1 DDX11L10 ZNF598 LOC107987233 MIR6859-4 NPRL3 LINC00235 UNKL FAM234A C16orf59 LOC105371045 LOC105371184 MPG SNHG9 LOC652276 PRR25 AXIN1 PRR35 PGP CASKIN1 TMEM204 PRSS27 PDPK1 FBXL16 TELO2 HAGHL WFIKKN1 TMEM8A CACNA1H HBA1 SYNGR3 MIR6767 NME3 NTHL1 IGFALS NARFL RNPS1 SOX8 NPW AMDHD2 EME2 SSTR5 NOXO1 ARHGDIG MIR3180-5 RPL3L ECI1 HN1L FAM173A MSRB1 WDR24 BAIAP3 MCRIP2 RHOT2 SNORD60 RGS11 MIR940 PTX4 SNRNP25 SNORA10 RNF151 E4F1 LOC100134368 RPUSD1 MLST8 MIR4516 LOC105371038 RAB26 RAB11FIP3 MIR3176 C16orf91 RHBDL1 HBM SNORA64 RPL23AP5 C1QTNF8 LMF1 LOC100287175 GNG13 LOC101929280 PKD1 LOC106660606 ERVK13-1 MIR3178 MEIOB POLR3K IFT140 SPSB3 MIR1225 TSR3 CCDC78 WASIR2 LOC105371049 MIR6511B1 HS3ST6 HBZ ABCA17P LOC105371046 TRAF7 MIR5587 JMJD8 PDIA2 KCTD5 RPS2 HBA2 TPSAB1 ABCA3 CEMP1 LINC00254 SLC9A3R2 NHLRC4 TBC1D24 MRPL28 TPSB2 ATP6V0C |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | MIR3677 DNASE1L2 NOXO1 PKD1 LOC106660606 GFER MIR3180-5 ZNF598 ECI1 MIR1225 SNORD60 LOC105371049 MIR6511B1 MIR940 TSC2 TRAF7 TBL3 PGP SNHG19 E4F1 CASKIN1 MLST8 BRICD5 MIR4717 MIR4516 ABCA3 SYNGR3 RAB26 SLC9A3R2 NTHL1 RNPS1 NPW |
nsv952901 | Chr.16:1888800 - 2135899 on Build GRCh38 | Deletion | SNORA64 SNORA78 TBL3 SNORA10 RNF151 NOXO1 PKD1 RPS2 GFER ZNF598 RPL3L MIR4516 MIR1225 MSRB1 SYNGR3 LOC105371049 MIR6511B1 SLC9A3R2 NTHL1 HS3ST6 TSC2 SNHG9 NPW NDUFB10 |
nsv1160329 | Chr.16:1961727 - 2128574 on Build GRCh38 | Deletion | SNORA64 SNORA78 TBL3 SNORA10 RNF151 NOXO1 PKD1 RPS2 GFER ZNF598 MIR1225 SYNGR3 LOC105371049 MIR6511B1 SLC9A3R2 NTHL1 TSC2 SNHG9 NPW NDUFB10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |