Assay Details
Target Gene Details
Entrez Gene ID: | 83986 |
Gene Name: | family with sequence similarity 234 member A |
Gene Aliases: |
C16orf9, ITFG3, gs19 |
Location: |
Chr.16:234546-269943 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM234A | NM_001284497.1 | NP_001271426.1 | ||
NM_032039.3 | NP_114428.1 | |||
NR_104317.1 | ||||
XM_005255622.1 | XP_005255679.1 | |||
XM_005255623.2 | XP_005255680.1 | |||
XM_006720957.1 | XP_006721020.1 | |||
XM_011522691.1 | XP_011520993.1 | |||
XM_011522692.1 | XP_011520994.1 | |||
XM_017023760.1 | XP_016879249.1 | |||
XM_017023761.1 | XP_016879250.1 | |||
XM_017023762.1 | XP_016879251.1 | |||
XM_017023763.1 | XP_016879252.1 | |||
XM_017023764.1 | XP_016879253.1 | |||
XM_017023765.1 | XP_016879254.1 | |||
AK057165.1 | BAB71374.1 | |||
AK294581.1 | ||||
AK310766.1 | ||||
AL136542.1 | CAB66477.1 | |||
CN262982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518291 | Chr.16:47354 - 254515 on Build GRCh38 | Loss | NPRL3 HBZ HBA1 FAM234A HBA2 HBM SNRNP25 LUC7L HBQ1 RHBDF1 MPG POLR3K |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MRPL28 TSR3 RPS2 LMF1-AS1 HAGHL PGP HBA2 MIR3178 PRR25 UNKL SNRNP25 TMEM8A MEIOB ECI1 CAPN15 LOC106660606 POLR3K SLC9A3R2 LOC107987233 TBL3 SNHG9 LOC101929280 LOC105371046 CHTF18 DNASE1L2 LOC100287175 RHBDF1 ZNF598 SNORD60 MIR3177 PRR35 CEMP1 DDX11L10 NME3 LOC105371184 TPSB2 RAB11FIP3 KCTD5 EME2 HS3ST6 GNG13 NHLRC4 FLJ42627 MIR4717 RAB26 MAPK8IP3 MIR3677 METTL26 WDR90 MRPS34 BAIAP3 MCRIP2 LUC7L AXIN1 MIR6768 MIR3180-5 RNF151 BRICD5 LOC652276 NTN3 NME4 WDR24 TMEM204 C16orf91 PTX4 MSRB1 NTHL1 LOC105371047 SNORA78 CCDC154 FBXL16 LOC105371045 LINC00254 MIR940 TBC1D24 HBZ LOC107984876 NOXO1 RPUSD1 TPSG1 PKD1 NDUFB10 GFER MIR6511B1 MIR4516 SSTR5-AS1 TRAF7 HAGH E4F1 STUB1 TPSAB1 MSLN SNHG19 ARHGDIG ABCA3 LINC00235 MIR6859-4 IGFALS NUBP2 CCNF SSTR5 MLST8 SPSB3 WASIR2 FAM234A PRSS29P LMF1 NARFL CCDC78 TELO2 LOC105371049 RNPS1 JMJD8 MIR5587 LOC729652 MPG MIR3176 PRSS27 FAHD1 FAM173A WFIKKN1 RHOT2 LOC105371038 HN1L HBQ1 MIR6767 AMDHD2 IFT140 CRAMP1 MIR662 HBM RHBDL1 SNORA64 C16orf59 ERVK13-1 CLCN7 DECR2 RPL3L PDIA2 UBE2I SNORA10 SOX8 HBA1 ABCA17P PIGQ ATP6V0C MIR1225 C1QTNF8 SYNGR3 GNPTG PDPK1 RGS11 NPRL3 NPW RAB40C RPL23AP5 CASKIN1 TSC2 LOC100134368 METRN TPSD1 CACNA1H |
esv3570031 | Chr.16:243389 - 278107 on Build GRCh38 | Gain | FAM234A RGS11 |
nsv1044142 | Chr.16:10765 - 255644 on Build GRCh38 | Gain | WASIR2 HBA1 FAM234A HBA2 SNRNP25 LUC7L HBQ1 DDX11L10 POLR3K MIR6859-4 NPRL3 HBZ HBM RHBDF1 MPG |
esv2667528 | Chr.16:246450 - 249976 on Build GRCh38 | Deletion | FAM234A |
esv3637560 | Chr.16:246387 - 250231 on Build GRCh38 | Loss | FAM234A |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | MRPL28 RAB11FIP3 FAM234A RPL23AP5 AXIN1 LOC100134368 TMEM8A ARHGDIG DECR2 PDIA2 RGS11 NME4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |