Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR3178 KCTD5 TSC2 MCRIP2 TPSB2 MPG NARFL HAGHL SNRNP25 MEIOB AXIN1 LOC105371049 ABCA17P LMF1-AS1 PRR35 NPW TSR3 CHTF18 LOC652276 HAGH SPSB3 LOC100134368 WASIR2 TRAF7 PGP C1QTNF8 NTHL1 RGS11 NUBP2 GFER RAB11FIP3 UBE2I NOXO1 TBL3 LOC107987233 MIR6859-4 ERVK13-1 CASKIN1 WDR90 MIR6767 SNHG9 NHLRC4 CCNF SNORA78 SSTR5-AS1 ECI1 HN1L SYNGR3 LOC105371047 MSRB1 LOC101929280 TPSD1 FBXL16 SNHG19 MIR940 MLST8 CCDC78 NME4 CCDC154 DNASE1L2 GNPTG STUB1 NTN3 CRAMP1 METTL26 SLC9A3R2 FAHD1 AMDHD2 MIR4516 RHOT2 CEMP1 SNORA10 NME3 ATP6V0C WDR24 FAM173A LINC00254 TELO2 BAIAP3 ZNF598 MIR6768 MIR6511B1 HBZ DDX11L10 SNORA64 JMJD8 MIR3176 PRR25 MRPS34 LUC7L SNORD60 PKD1 ABCA3 PDIA2 RNPS1 MSLN MIR3677 BRICD5 MRPL28 CACNA1H HBA2 RPS2 RAB40C C16orf91 LOC106660606 TPSG1 LOC107984876 UNKL E4F1 HBA1 METRN PTX4 RHBDL1 HBM LOC105371046 SSTR5 MIR4717 RPUSD1 TBC1D24 LOC105371045 LOC105371184 HS3ST6 LOC105371038 MIR5587 LMF1 RAB26 NDUFB10 HBQ1 TMEM204 MAPK8IP3 ARHGDIG MIR1225 TMEM8A GNG13 WFIKKN1 C16orf59 FAM234A CLCN7 IGFALS RPL23AP5 PRSS27 LOC100287175 MIR3180-5 RPL3L PRSS29P IFT140 TPSAB1 PDPK1 RNF151 NPRL3 EME2 POLR3K CAPN15 MIR3177 LOC729652 DECR2 SOX8 FLJ42627 RHBDF1 MIR662 LINC00235 PIGQ |
dgv4810n54 | Chr.16:380011 - 445662 on Build GRCh38 | Loss | RPL23AP5 DECR2 NME4 LOC100134368 TMEM8A RAB11FIP3 |
nsv9320 | Chr.16:427245 - 432556 on Build GRCh38 | Loss | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | FAM234A RPL23AP5 DECR2 NME4 ARHGDIG MRPL28 LOC100134368 PDIA2 TMEM8A RAB11FIP3 RGS11 AXIN1 |
esv3637574 | Chr.16:427856 - 430795 on Build GRCh38 | Loss | RAB11FIP3 |
nsv570956 | Chr.16:376432 - 437728 on Build GRCh38 | Gain | RPL23AP5 DECR2 NME4 LOC100134368 TMEM8A RAB11FIP3 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | CAPN15 RPL23AP5 DECR2 NME4 MRPL28 LOC100134368 TMEM8A LINC00235 RAB11FIP3 AXIN1 |
nsv457303 | Chr.16:383623 - 445662 on Build GRCh38 | Loss | RPL23AP5 DECR2 NME4 LOC100134368 RAB11FIP3 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | RPL23AP5 DECR2 NME4 MRPL28 LOC100134368 TMEM8A RAB11FIP3 AXIN1 |
esv2660167 | Chr.16:427853 - 430781 on Build GRCh38 | Deletion | RAB11FIP3 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | FAM234A RPL23AP5 DECR2 NME4 ARHGDIG MRPL28 LOC100134368 PDIA2 TMEM8A RAB11FIP3 RGS11 AXIN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |