Assay Details
Target Gene Details
Entrez Gene ID: | 22980 |
Gene Name: | transcription factor 25 |
Gene Aliases: |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
Location: |
Chr.16:89873583-89911384 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573744 | Chr.16:89738447 - 89915950 on Build GRCh38 | Loss | LOC105371419 ZNF276 FANCA SPIRE2 TCF25 LOC107984817 |
esv2758671 | Chr.16:89804324 - 89901541 on Build GRCh38 | Loss | LOC105371419 FANCA SPIRE2 TCF25 LOC107984817 |
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | LOC105371419 TUBB3 DEF8 DBNDD1 VPS9D1-AS1 TCF25 LOC107984817 CDK10 SPATA2L SPATA33 AFG3L1P ZNF276 MC1R GAS8 FANCA CENPBD1 SPIRE2 CHMP1A GAS8-AS1 DPEP1 VPS9D1 |
nsv573810 | Chr.16:89857498 - 90037251 on Build GRCh38 | Loss | TUBB3 AFG3L1P DEF8 DBNDD1 MC1R GAS8 CENPBD1 SPIRE2 GAS8-AS1 TCF25 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | TUBB3 DEF8 PRDM7 LOC105371414 LOC107984817 RPL13 URAHP ANKRD11 GAS8 CHMP1A LOC101927817 DPEP1 LOC105371419 FAM157C LOC100287036 TUBB8P7 DBNDD1 VPS9D1-AS1 TCF25 CDK10 SPATA2L SPATA33 AFG3L1P ZNF778 CPNE7 ZNF276 MC1R FANCA CENPBD1 SPIRE2 GAS8-AS1 SPG7 VPS9D1 SNORD68 |
nsv573799 | Chr.16:89852186 - 90022379 on Build GRCh38 | Gain | LOC105371419 TUBB3 AFG3L1P DEF8 DBNDD1 MC1R GAS8 CENPBD1 SPIRE2 TCF25 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC107987238 MVD DEF8 PRDM7 LOC105371414 PIEZO1 RPL13 URAHP ACSF3 CDH15 CBFA2T3 GAS8 CHMP1A DPEP1 LOC105371409 LOC105371419 CYBA FAM157C LOC100287036 LOC100289580 VPS9D1-AS1 LOC339059 TCF25 CDK10 SPATA2L SPATA33 AFG3L1P ZNF778 ZNF276 GALNS MC1R CENPBD1 GAS8-AS1 SPG7 SNORD68 RNF166 TUBB3 CDT1 SLC22A31 LOC107984817 LOC400558 LOC100129697 LOC101927793 IL17C ANKRD11 LOC101927817 SNAI3 TUBB8P7 DBNDD1 TRAPPC2L CTU2 PABPN1L APRT LINC00304 CPNE7 FANCA SPIRE2 MIR4722 VPS9D1 SNAI3-AS1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |