Assay Details
Target Gene Details
Entrez Gene ID: | 81889 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 1 |
Gene Aliases: |
C16orf36, YISKL |
Location: |
Chr.16:1827224-1840207 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAHD1 | NM_001018104.2 | NP_001018114.1 | ||
NM_001142398.1 | NP_001135870.1 | |||
BC063017.1 | AAH63017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss | EME2 MAPK8IP3 SPSB3 FAHD1 MIR3177 HAGH MEIOB NUBP2 IGFALS MRPS34 NME3 |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | FAHD1 HAGH MEIOB |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | FAHD1 HAGH MEIOB |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | LOC105371046 MIR4516 HS3ST6 WFIKKN1 PRSS27 TBC1D24 UNKL MIR4717 MIR3178 WDR24 MIR1225 WASIR2 MIR3677 ABCA17P RPL3L PRSS29P MIR6859-4 METTL26 TSC2 MIR662 ZNF598 ABCA3 HBA2 PRR35 NME3 RPL23AP5 DNASE1L2 RAB26 MIR3180-5 MRPL28 FBXL16 RPUSD1 NDUFB10 ERVK13-1 FAHD1 NME4 LOC729652 SNRNP25 NARFL CCDC154 AMDHD2 SNORA10 MSLN HBM TSR3 ATP6V0C LOC105371045 SNORD60 MIR6768 MIR3177 TELO2 MCRIP2 LOC105371047 SSTR5-AS1 GNG13 FLJ42627 PGP TRAF7 PTX4 TMEM8A BAIAP3 LOC652276 SPSB3 RHBDL1 PKD1 MIR3176 HAGH TBL3 PRR25 IFT140 CCDC78 SLC9A3R2 HBA1 STUB1 IGFALS MRPS34 MAPK8IP3 PDPK1 WDR90 CHTF18 ECI1 PIGQ MEIOB LOC106660606 LINC00235 KCTD5 RGS11 LOC105371038 CEMP1 RHBDF1 LOC100287175 E4F1 MIR940 GNPTG SNHG9 AXIN1 HN1L PDIA2 CRAMP1 RNPS1 SOX8 TPSB2 DDX11L10 MIR5587 MLST8 NTHL1 C1QTNF8 SYNGR3 NOXO1 LUC7L METRN MIR6767 FAM234A NUBP2 GFER FAM173A LOC107984876 RAB11FIP3 LOC107987233 LMF1 CASKIN1 TPSAB1 EME2 CCNF LMF1-AS1 BRICD5 TPSG1 MSRB1 RNF151 LOC100134368 LOC105371184 NPW TPSD1 MIR6511B1 LINC00254 POLR3K HBQ1 C16orf59 TMEM204 HBZ UBE2I CAPN15 CLCN7 C16orf91 RHOT2 DECR2 NTN3 RPS2 SNORA78 SNORA64 SNHG19 CACNA1H SSTR5 MPG LOC105371049 HAGHL LOC101929280 JMJD8 NHLRC4 NPRL3 ARHGDIG RAB40C |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | LOC105371046 TMEM204 IFT140 MIR3177 TELO2 LOC105371047 NUBP2 IGFALS MRPS34 NME3 EME2 MAPK8IP3 SPSB3 FAHD1 HAGH HN1L CRAMP1 MEIOB |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | FAHD1 HAGH MEIOB NUBP2 IGFALS |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | MIR3177 NUBP2 IGFALS MRPS34 NME3 EME2 MAPK8IP3 SPSB3 FAHD1 HAGH HN1L CRAMP1 MEIOB |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | FAHD1 HAGH MEIOB |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | LINC00254 FAHD1 HAGH MEIOB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |