Assay Details
Target Gene Details
Entrez Gene ID: | 1428 |
Gene Name: | crystallin mu |
Gene Aliases: |
DFNA40, THBP |
Location: |
Chr.16:21258518-21303136 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYM | NM_001888.4 | NP_001879.1 | ||
BG033512.1 | ||||
BI860684.1 | ||||
BX648477.1 | CAI46030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34235 | Chr.16:21276264 - 21986230 on Build GRCh38 | Gain | SLC7A5P2 RRN3P1 SNX29P1 NPIPB4 UQCRC2 CRYM-AS1 NPIPB3 METTL9 LOC101927814 PDZD9 LOC100190986 OTOA CRYM SMG1P3 MIR3680-1 IGSF6 |
nsv571623 | Chr.16:21084049 - 21341186 on Build GRCh38 | Gain | ZP2 ANKS4B CRYM DNAH3 CRYM-AS1 TMEM159 |
esv3638164 | Chr.16:21185596 - 21338776 on Build GRCh38 | Gain | ZP2 ANKS4B CRYM CRYM-AS1 |
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | DCUN1D3 GDE1 ACSM2B KNOP1 ITPRIPL2 LOC81691 CLEC19A ACSM5 GPRC5B UMOD ACSM3 ACSM1 ACSM2A COQ7 LOC100190986 PDILT ZP2 SMG1P3 DNAH3 LOC102723385 TMEM159 C16orf62 SNX29P1 ANKS4B GPR139 SYT17 IQCK CRYM-AS1 NPIPB3 CCP110 ERI2 LYRM1 CRYM TMC7 GP2 THUMPD1 TMC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |