Assay Details
Target Gene Details
Entrez Gene ID: | 54620 |
Gene Name: | F-box and leucine rich repeat protein 19 |
Gene Aliases: |
CXXC11, Fbl19, JHDM1C |
Location: |
Chr.16:30923055-30948783 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXL19 | NM_001099784.2 | NP_001093254.2 | ||
NM_001282351.1 | NP_001269280.1 | |||
AK098777.1 | BAC05410.1 | |||
AK127701.1 | ||||
AK302669.1 | ||||
AL137589.1 | CAB70830.1 | |||
BC059173.1 | AAH59173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv457483 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss | ZNF668 ZNF646 BCL7C LOC101928762 MIR762 ORAI3 SETD1A PRSS8 MIR762HG STX1B KAT8 FBXL19 FUS STX4 PRSS53 BCKDK FBXL19-AS1 PRSS36 VKORC1 CTF1 HSD3B7 |
dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss | STX4 BCL7C MIR762 ORAI3 SETD1A MIR762HG STX1B FBXL19-AS1 FBXL19 CTF1 HSD3B7 MIR4519 |
nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion | BCL7C MIR762 ORAI3 SETD1A MIR762HG STX1B FBXL19-AS1 FBXL19 CTF1 HSD3B7 |
nsv833189 | Chr.16:30808387 - 30949474 on Build GRCh38 | Loss | BCL7C MIR762 ORAI3 MIR762HG FBXL19-AS1 FBXL19 CTF1 MIR4519 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |