Assay Details
Target Gene Details
Entrez Gene ID: | 84656 |
Gene Name: | glyoxylate reductase 1 homolog |
Gene Aliases: |
BM045, HIBDL, N-PAC, NP60 |
Location: |
Chr.16:4803203-4847388 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26756 | Chr.16:4772529 - 4855231 on Build GRCh38 | Gain+Loss | SMIM22 UBN1 SEPT12 ROGDI GLYR1 |
esv2422516 | Chr.16:4775920 - 5160979 on Build GRCh38 | Deletion | EEF2KMT PPL SMIM22 UBN1 NAGPA-AS1 SEPT12 ALG1 ROGDI NAGPA SEC14L5 C16orf89 GLYR1 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | GLIS2 SMIM22 HMOX2 UBN1 ANKS3 SEC14L5 C16orf96 EEF2KMT TFAP4 NAGPA-AS1 NUDT16L1 UBALD1 ROGDI LOC105371063 MGRN1 ADCY9 LINC01569 NMRAL1 PPL ZNF500 GLIS2-AS1 MIR6769A CORO7 CDIP1 NAGPA GLYR1 RBFOX1 PAM16 SRL CORO7-PAM16 VASN SEPT12 ALG1 DNAJA3 C16orf71 C16orf89 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |