Assay Details
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | NP_001157032.1 | ||
NM_152764.2 | NP_689977.2 | |||
BC029829.1 | AAH29829.1 | |||
CD300613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss | FAHD1 MEIOB LINC00254 HS3ST6 |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PKD1 ARHGDIG UNKL HBA1 RPUSD1 TRAF7 PDIA2 EME2 LMF1 GFER LOC105371049 MIR3677 LUC7L DECR2 SSTR5 LOC107987233 SPSB3 SSTR5-AS1 DNASE1L2 DDX11L10 SOX8 MPG GNPTG LOC105371038 RHBDF1 WASIR2 CACNA1H ABCA17P RAB40C NPW LOC106660606 NHLRC4 RGS11 CLCN7 MIR6768 TPSG1 ECI1 NARFL SNORA78 AMDHD2 C16orf59 LMF1-AS1 RPS2 MSRB1 CCDC154 METTL26 RHOT2 ATP6V0C ABCA3 TBL3 RAB11FIP3 PTX4 HS3ST6 MIR662 FAHD1 LOC100134368 TELO2 HBA2 CASKIN1 FAM234A PRSS29P PIGQ CCNF MIR5587 WDR90 MIR4717 HBQ1 CCDC78 LINC00235 HBM C1QTNF8 MIR1225 LOC101929280 MIR940 TPSB2 SNORD60 RHBDL1 SLC9A3R2 JMJD8 FAM173A NTN3 MSLN LOC100287175 LOC652276 MRPL28 NME3 NUBP2 HAGH IFT140 MIR3177 TSC2 NDUFB10 MIR3180-5 C16orf91 WFIKKN1 NPRL3 RPL23AP5 LOC107984876 MIR6511B1 CAPN15 E4F1 LOC105371046 IGFALS RAB26 NME4 LOC105371184 ERVK13-1 WDR24 SNRNP25 LOC729652 NOXO1 BRICD5 TMEM204 RPL3L MLST8 POLR3K SNORA64 ZNF598 MIR4516 RNPS1 KCTD5 MCRIP2 BAIAP3 SNHG19 SNHG9 METRN CHTF18 TPSAB1 TMEM8A GNG13 LOC105371047 CRAMP1 FBXL16 TBC1D24 MIR3176 LINC00254 HBZ PRSS27 AXIN1 CEMP1 SNORA10 NTHL1 PRR25 TSR3 MIR3178 MIR6859-4 FLJ42627 TPSD1 SYNGR3 MIR6767 RNF151 LOC105371045 MEIOB HAGHL STUB1 HN1L MAPK8IP3 PDPK1 PGP UBE2I PRR35 MRPS34 |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | FAHD1 MEIOB HAGH |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | FAHD1 LOC105371046 TELO2 IGFALS EME2 MEIOB NME3 NUBP2 HAGH LOC105371047 HN1L CRAMP1 MAPK8IP3 IFT140 MIR3177 SPSB3 TMEM204 MRPS34 |
nsv1046474 | Chr.16:1846666 - 1873690 on Build GRCh38 | Loss | MEIOB |
nsv519366 | Chr.16:1851210 - 1961125 on Build GRCh38 | Loss | MEIOB LINC00254 NDUFB10 RPL3L HS3ST6 MSRB1 |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | FAHD1 MEIOB NUBP2 HAGH IGFALS |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | FAHD1 MEIOB LINC00254 HAGH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |